Search
Search
| Gene Symbol: Dok2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106048.1 | NP_001099518.1 | 2 - 3 | 369 | 76 | |
| XM_006252262.1 | XP_006252324.1 | 3 - 4 | 1138 | 76 | ||
| XM_006252264.2 | XP_006252326.1 | 3 - 4 | 1599 | 76 | ||
| XM_008770826.1 | XP_008769048.1 | 2 - 3 | 2573 | 76 | ||
Set Membership: |
|