Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ? 立即註冊
  • 產品
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Greener Products
    • 查看所有產品類別
  • 應用
    • 生物製程
    • 細胞培養和轉染
    • 食品和飲料
    • 實驗室解決方案
    • 製藥和生物製藥
    • 即時定量PCR
    • 半導體分析
    • 臨床和診斷
    • 數位解決方案
    • 分子光譜、同位素與元素分析
    • 查看所有應用與技術
  • 服務
    • 360° CDMO 和 CRO 服務
    • CDMO 服务
    • 临床试验 CRO 服务
    • 客製化服務
    • 金融與租賃服務
    • 儀器服務
    • 實驗室資訊學
    • 授權與商業供應
    • 培訓服務
    • 查看所有服務
  • 支援
    • 線上訂購協助
    • 數位解決方案
    • 產品支持
    • 技術相關資訊
    • 培訓與教育
    • 查看所有協助與支援
  • 熱門商品
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • 我們的服務對象
    • 生物技術
    • 生物製藥
    • CDMO
    • 實驗室診斷
    • 工業與應用科學
  • 特價商品
  • 聯絡我們
  • 快速訂購
  • 訂單狀態與貨物追蹤
  • 文件和證書
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • 訂單查詢
          • 快速訂購
          • 登入
            登入
            還沒有帳戶嗎 ? 立即註冊
            • 帳戶
            • 訂單
            • Connect: Lab, Data, Apps
            • 客製化產品與項目
            • 儀器管理
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01467966_m1
          See other TM9SF2 GE Assays ›
          Gene Symbol:
          Tm9sf2
          Gene Name:
          transmembrane 9 superfamily member 2
          Gene Aliases:
          -
          Chromosome Location:
          Chr.15: 108526341 - 108579268 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01467966_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tm9sf2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001005554.1 NP_001005554.1 16 - 17 2087 107
          GenBank mRNA BC081873.1 - 16 - 17 2087 7107003 107
          FQ209454.1 - 4 - 5 515 107
          FQ226287.1 - 1 - 2 185 107

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1500001199.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Tm9sf2

          Panther Classification:

          Molecular Function -

          transporter

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P398399 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 382282 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 760368 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 120532 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 162571 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 148842 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 802518 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 273756 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 398996 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 226656 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 840814 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 565115 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 812237 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 543991 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 911684 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 69503 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 627566 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 710286 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 68148 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 785323 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 478704 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 845543 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 225442 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 209834 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 621685 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 750265 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 259856 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 290081 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 655955 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 946980 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 549756 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 180764 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 469394 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 742585 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 262399 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 644593 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 552953 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 12596 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 268861 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 33932 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 85158 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 49403 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 1855 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 17659 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P398399 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P398399 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P398399 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P398399 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 15726 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 64886 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 99304 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 52451 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10782140 - -
          Affymetrix Rat Gene 1.0 ST Array 10782132 - -
          Affymetrix Rat Gene 1.0 ST Array 10782130 - -
          Affymetrix Rat Gene 1.0 ST Array 10782134 - -
          Affymetrix Rat Gene 1.0 ST Array 10782135 - -
          Affymetrix Rat Gene 1.0 ST Array 10782131 - -
          Affymetrix Rat Gene 1.0 ST Array 10782143 - -
          Affymetrix Rat Gene 1.0 ST Array 10782128 - -
          Affymetrix Rat Gene 1.0 ST Array 10782139 - -
          Affymetrix Rat Gene 1.0 ST Array 10782142 - -
          Affymetrix Rat Gene 1.0 ST Array 10782133 - -
          Affymetrix Rat Gene 1.0 ST Array 10782136 - -
          Affymetrix Rat Gene 1.0 ST Array 10782137 - -
          Affymetrix Rat Gene 1.0 ST Array 10782127 - -
          Affymetrix Rat Gene 1.0 ST Array 10782138 - -
          Affymetrix Rat Gene 1.0 ST Array 10782129 - -
          Affymetrix Rat Gene 1.0 ST Array 10782141 - -
          Affymetrix Rat Gene 1.1 ST Array 10782126 - -
          Rat Genome 230 2.0 Array 1367496_at - -
          Rat Genome 230 2.0 Array 1367496_PM_at - -
          Affymetrix HT Rat Focus Array 1367496_PM_at - -
          Affymetrix HT Rat Focus Array 1367496_at - -
          Affymetrix Rat Clariom S Assay PSR1500007522.rn.1 TC1500001199.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1500007521.rn.1 TC1500001199.rn.1 RTA TC
          Rat Expression Set 230 1367496_PM_at - -
          Rat Expression Set 230 1367496_at - -
          Rat Expression Set 230 1367496_at - -
          Rat Expression Set 230 1367496_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10782126 - -
          Rat Genome U34 Set rc_AI179901_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22052505 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1351439 - -
          RatRef-12 Expression BeadChip ILMN_67476 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          產品訂購 Plus Icon Minus Icon
          • 訂單狀態
          • 訂購協助
          • 快速訂購
          • 現貨供應中心
          • 電子採購
          支持 Plus Icon Minus Icon
          • 協助與支持
          • 聯絡我們
          • 技術支持中心
          • 搜尋文件和證書
          • 回報網站問題
          資源 Plus Icon Minus Icon
          • 學習中心
          • 促銷活動
          • 活動和網路研討會
          • 社群媒體
          關於 Thermo Fisher Plus Icon Minus Icon
          • 關於我們 關於我們
          • 人才招募 人才招募
          • 投資者資訊 投資者資訊
          • 新聞 新聞
          • 企業社會責任 企業社會責任
          • 商標
          • 政策和聲明
          品牌 Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline