Search
Search
| Gene Symbol: R3hcc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006252286.2 | XP_006252348.1 | 5 - 6 | 1495 | 65 | |
| XM_006252288.2 | XP_006252350.1 | 5 - 6 | 1498 | 65 | ||
| XM_006252289.1 | XP_006252351.1 | 5 - 6 | 1284 | 65 | ||
Set Membership: |
|