Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01468046_m1
          See other PDLIM2 GE Assays ›
          Gene Symbol:
          Pdlim2
          Gene Name:
          PDZ and LIM domain 2
          Gene Aliases:
          -
          Chromosome Location:
          Chr.15: 51841508 - 51856785 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01468046_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Pdlim2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001007622.1 NP_001007623.1 9 - 10 996 75
          XM_006252255.2 XP_006252317.1 9 - 10 1011 75
          XM_006252256.2 XP_006252318.1 9 - 10 1006 75
          XM_006252257.2 XP_006252319.1 9 - 10 1528 75
          XM_006252258.1 XP_006252320.1 9 - 10 896 75
          XM_006252259.2 XP_006252321.1 9 - 10 1563 75
          XM_008770823.1 XP_008769045.1 9 - 10 897 75
          GenBank mRNA AY531526.1 - 9 - 10 978 75
          BC079091.1 - 9 - 10 996 7123600 75

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1500002483.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Pdlim2

          Panther Classification:

          Molecular Function -

          actin or actin-binding cytoskeletal protein

          Gene Ontology Categories:

          Process(es)

          protein binding
          zinc ion binding
          filamin binding
          myosin heavy chain binding
          muscle alpha-actinin binding
          alpha-actinin binding

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P106456 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 722159 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 840000 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 849322 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 886013 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 292409 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 824717 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 537605 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 246624 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 380009 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 127660 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P106456 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P106456 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P106456 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P17609 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P106456 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 104178 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 101829 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10784975 - -
          Affymetrix Rat Gene 1.0 ST Array 10784971 - -
          Affymetrix Rat Gene 1.0 ST Array 10784976 - -
          Affymetrix Rat Gene 1.0 ST Array 10784979 - -
          Affymetrix Rat Gene 1.0 ST Array 10784973 - -
          Affymetrix Rat Gene 1.0 ST Array 10784974 - -
          Affymetrix Rat Gene 1.0 ST Array 10784972 - -
          Affymetrix Rat Gene 1.0 ST Array 10784977 - -
          Affymetrix Rat Gene 1.0 ST Array 10784978 - -
          Affymetrix Rat Gene 1.1 ST Array 10784968 - -
          Rat Genome 230 2.0 Array 1375367_at - -
          Rat Genome 230 2.0 Array 1375367_PM_at - -
          Affymetrix HT Rat Focus Array 1375367_at - -
          Affymetrix HT Rat Focus Array 1375367_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1500016917.rn.1 TC1500002483.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1500016918.rn.1 TC1500002483.rn.1 RTA TC
          Rat Expression Set 230 1375367_at - -
          Rat Expression Set 230 1375367_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10784968 - -
          Rat Genome U34 Set rc_AA893726_at - -
          Rat Genome U34 Set rc_AI170340_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20999913 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline