Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01468067_m1
          See other RHOBTB2 GE Assays ›
          Gene Symbol:
          Rhobtb2
          Gene Name:
          Rho-related BTB domain containing 2
          Gene Aliases:
          -
          Chromosome Location:
          Chr.15: 51467272 - 51485562 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01468067_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rhobtb2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001013133.1 NP_001013151.1 7 - 8 2138 61
          GenBank mRNA BC091309.1 - 7 - 8 2138 7311705 61

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1500001979.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Rattus norvegicus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Rhobtb2

          Panther Classification:

          Molecular Function -

          small GTPase

          Gene Ontology Categories:

          Function(s) Process(es)

          small GTPase mediated signal transduction
          GTP binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P470976 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 599088 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 277656 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 318 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 756071 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 367771 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 845142 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 375484 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 334443 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 26023 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 767918 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 897155 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 897735 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 328532 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 19490 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 317418 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 132406 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 43400 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 163669 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P470976 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P470976 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P470976 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P470976 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 36464 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 90564 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 78804 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10784930 - -
          Affymetrix Rat Gene 1.0 ST Array 10784928 - -
          Affymetrix Rat Gene 1.0 ST Array 10784931 - -
          Affymetrix Rat Gene 1.0 ST Array 10784937 - -
          Affymetrix Rat Gene 1.0 ST Array 10784934 - -
          Affymetrix Rat Gene 1.0 ST Array 10784935 - -
          Affymetrix Rat Gene 1.0 ST Array 10784932 - -
          Affymetrix Rat Gene 1.0 ST Array 10784936 - -
          Affymetrix Rat Gene 1.0 ST Array 10784933 - -
          Affymetrix Rat Gene 1.0 ST Array 10784929 - -
          Affymetrix Rat Gene 1.1 ST Array 10784927 - -
          Rat Genome 230 2.0 Array 1390001_PM_at - -
          Rat Genome 230 2.0 Array 1382279_PM_at - -
          Rat Genome 230 2.0 Array 1382279_at - -
          Rat Genome 230 2.0 Array 1390001_at - -
          Affymetrix HT Rat Focus Array 1382279_PM_at - -
          Affymetrix HT Rat Focus Array 1390001_PM_at - -
          Affymetrix HT Rat Focus Array 1382279_at - -
          Affymetrix HT Rat Focus Array 1390001_at - -
          Affymetrix Rat Clariom S Assay PSR1500013454.rn.1 TC1500001979.rn.1 RTA TC
          Rat Expression Set 230 1390001_at - -
          Rat Expression Set 230 1390001_PM_at - -
          Rat Expression Set 230 1382279_PM_at - -
          Rat Expression Set 230 1382279_at - -
          Affymetrix Rat Gene 1.0 ST Array 10784927 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1353915 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline