Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01468126_m1
          See other DOCK9 GE Assays ›
          Gene Symbol:
          Dock9
          Gene Name:
          dedicator of cytokinesis 9
          Gene Aliases:
          Trg
          Chromosome Location:
          Chr.15: 107945664 - 108210985 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01468126_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 4,430.00
          Your Price
          Online offer:
          4430.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Dock9
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001105759.2 NP_001099229.2 30 - 31 3405 74
          XM_006252481.2 XP_006252543.1 30 - 31 3516 74
          XM_006252488.2 XP_006252550.1 30 - 31 3516 74
          XM_006252490.2 XP_006252552.1 30 - 31 3516 74
          XM_006252493.2 XP_006252555.1 30 - 31 3971 74
          XM_006252496.2 XP_006252558.1 30 - 31 3540 74
          XM_006252499.2 XP_006252561.1 30 - 31 3515 74
          XM_006252501.2 XP_006252563.1 30 - 31 3515 74
          XM_006252502.2 XP_006252564.1 30 - 31 3515 74
          XM_006252503.2 XP_006252565.1 30 - 31 3564 74
          XM_006252505.2 XP_006252567.1 30 - 31 3540 74
          XM_008770951.1 XP_008769173.1 30 - 31 3516 74
          XM_008770952.1 XP_008769174.1 30 - 31 3516 74
          XM_008770953.1 XP_008769175.1 30 - 31 3516 74
          XM_008770954.1 XP_008769176.1 30 - 31 3516 74
          XM_008770955.1 XP_008769177.1 30 - 31 3541 74
          XM_008770956.1 XP_008769178.1 30 - 31 3541 74
          XM_008770957.1 XP_008769179.1 30 - 31 3516 74
          XM_008770958.1 XP_008769180.1 30 - 31 3516 74
          XM_008770959.1 XP_008769181.1 30 - 31 3516 74
          XM_008770960.1 XP_008769182.1 30 - 31 3516 74
          XM_008770961.1 XP_008769183.1 30 - 31 3516 74
          XM_008770962.1 XP_008769184.1 30 - 31 3516 74
          XM_008770963.1 XP_008769185.1 30 - 31 3516 74
          XM_008770964.1 XP_008769186.1 30 - 31 3581 74
          XM_008770965.1 XP_008769187.1 30 - 31 3516 74
          XM_008770966.1 XP_008769188.1 30 - 31 3516 74
          XM_008770967.1 XP_008769189.1 30 - 31 3515 74
          XM_008770968.1 XP_008769190.1 30 - 31 3976 74
          XM_008770969.1 XP_008769191.1 30 - 31 3977 74

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1500002429.rn.1

          Set Membership:

          Amplicon length between 71 and 85 Recommended Assay Rattus norvegicus Probe spans exons

          Panther Classification:

          Molecular Function -

          guanyl-nucleotide exchange factor

          Gene Ontology Categories:

          Function(s) Process(es)

          small GTPase mediated signal transduction
          positive regulation of GTPase activity
          guanyl-nucleotide exchange factor activity
          Rho guanyl-nucleotide exchange factor activity
          Rho GTPase binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Clariom S Assay PSR1500016302.rn.1 TC1500002429.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1500016295.rn.1 TC1500002429.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1500016298.rn.1 TC1500002429.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1500016303.rn.1 TC1500002429.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1500016304.rn.1 TC1500002429.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1500016296.rn.1 TC1500002429.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1500016297.rn.1 TC1500002429.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1500016299.rn.1 TC1500002429.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1500016301.rn.1 TC1500002429.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1500016300.rn.1 TC1500002429.rn.1 RTA TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline