Search
Search
| Gene Symbol: Akap11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_012773.2 | NP_036905.2 | 2 - 3 | 187 | 83 | |
| XM_006252344.1 | XP_006252406.1 | 3 - 4 | 361 | 83 | ||
| XM_008770886.1 | XP_008769108.1 | 3 - 4 | 1253 | 83 | ||
| XM_008770887.1 | XP_008769109.1 | 3 - 4 | 443 | 83 | ||
| XM_008770888.1 | XP_008769110.1 | 3 - 4 | 443 | 83 | ||
| XM_008770889.1 | XP_008769111.1 | 3 - 4 | 443 | 83 | ||
| XM_008770890.1 | XP_008769112.1 | 3 - 4 | 443 | 83 | ||
Set Membership: |
|