Search
Search
| Gene Symbol: Hectd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006240086.1 | XP_006240148.1 | 27 - 28 | 5260 | 61 | |
| XM_006240087.1 | XP_006240149.1 | 27 - 28 | 5254 | 61 | ||
| XM_006240088.1 | XP_006240150.1 | 27 - 28 | 5119 | 61 | ||
| XM_006240089.1 | XP_006240151.1 | 27 - 28 | 5260 | 61 | ||
| GenBank mRNA | FQ211662.1 | - | 3 - 4 | 335 | 61 | |
Set Membership: |
|