Search
Search
| Gene Symbol: Sav1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001097581.1 | NP_001091050.1 | 1 - 2 | 264 | 71 | |
| GenBank mRNA | AM261215.1 | - | 1 - 2 | 264 | 71 | |
| BC161803.1 | - | 1 - 2 | 111 | 8371277 | 71 | |
Set Membership: |
|