Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01468735_m1
          See other TSSC1 GE Assays ›
          Gene Symbol:
          Tssc1
          Gene Name:
          tumor suppressing subtransferable candidate 1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.6: 48021771 - 48135241 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01468735_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tssc1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001012192.1 NP_001012192.1 8 - 9 1058 63
          GenBank mRNA BC087633.1 - 8 - 9 1058 7320156 63

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0600000540.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Tssc1

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P053943 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 839628 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 257259 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 268608 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 874619 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 357058 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 827810 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 576356 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 615514 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 190779 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 618951 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 701735 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 620232 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 218142 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 50239 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 681725 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 383606 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 4912 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 359392 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 721166 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 493281 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 95631 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 679531 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 921351 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 568290 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 321846 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 667796 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 373877 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 54128 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 349413 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 43028 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 478665 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 783883 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 184847 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 799456 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 737114 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 231845 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 406129 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 581236 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 714502 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 674867 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 181511 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 252801 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 319089 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 216195 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 72154 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 956611 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 718829 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 647952 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 127160 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 78209 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 522943 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 968219 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 9585 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 725161 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 661715 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 632429 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 598243 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 962398 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 710574 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 64519 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 86801 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 519501 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 287980 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 787144 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 7075 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 161277 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 534587 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 888917 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 455832 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 835680 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 590017 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 1895 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 380574 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 654483 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 628854 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 958970 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 284435 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 931480 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 701550 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 918949 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 822019 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 86482 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 497210 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 450112 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 661246 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 45646 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 161373 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 96471 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 33345 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 17105 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 13787 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 91325 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 101377 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 90000 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 85131 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 145073 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 106275 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P053943 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P053943 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P053943 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P499433 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 66404 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 96670 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 25977 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 3298 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 58618 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 12572 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 104901 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10883990 - -
          Affymetrix Rat Gene 1.0 ST Array 10883993 - -
          Affymetrix Rat Gene 1.0 ST Array 10883992 - -
          Affymetrix Rat Gene 1.0 ST Array 10883998 - -
          Affymetrix Rat Gene 1.0 ST Array 10884000 - -
          Affymetrix Rat Gene 1.0 ST Array 10883997 - -
          Affymetrix Rat Gene 1.0 ST Array 10883996 - -
          Affymetrix Rat Gene 1.0 ST Array 10883988 - -
          Affymetrix Rat Gene 1.0 ST Array 10883989 - -
          Affymetrix Rat Gene 1.0 ST Array 10883991 - -
          Affymetrix Rat Gene 1.0 ST Array 10883999 - -
          Affymetrix Rat Gene 1.1 ST Array 10883987 - -
          Rat Genome 230 2.0 Array 1379385_at - -
          Rat Genome 230 2.0 Array 1379385_PM_at - -
          Affymetrix HT Rat Focus Array 1379385_at - -
          Affymetrix HT Rat Focus Array 1379385_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0600003815.rn.1 TC0600000540.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0600003814.rn.1 TC0600000540.rn.1 RTA TC
          Rat Expression Set 230 1379385_at - -
          Rat Expression Set 230 1379385_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10883987 - -
          Rat Genome U34 Set rc_AI103178_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20978664 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1376645 - -
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1375485 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-green-7b6576f7bc-knt4q:80/100.66.75.14:80.
          git-commit: 6d309ebe7a31b2ceadeb87f43b473eebab110cf3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-2026.03.11-1.0