Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01468962_m1
          See other ATL1 GE Assays ›
          Gene Symbol:
          Atl1
          Gene Name:
          atlastin GTPase 1
          Gene Aliases:
          Spg3a
          Chromosome Location:
          Chr.6: 92229785 - 92370428 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01468962_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Atl1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001009831.2 NP_001009831.1 8 - 9 942 69
          XM_006240177.1 XP_006240239.1 7 - 8 878 69
          GenBank mRNA AY581896.2 - 8 - 9 942 69
          BC092645.1 - 7 - 8 865 7318365 69

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0600000906.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Atl1

          Panther Classification:

          Molecular Function -

          heterotrimeric G-protein

          Gene Ontology Categories:

          Function(s) Process(es)

          endoplasmic reticulum organization
          axonogenesis
          metabolic process
          protein homooligomerization
          GTPase activity
          protein binding
          GTP binding
          identical protein binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P084513 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 834433 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 519 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 494599 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 452162 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 957485 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 887949 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 169720 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 854937 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 90103 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 96757 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 280265 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 13392 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 927633 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 457119 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 908224 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 266495 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 483831 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 147018 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 746744 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 403956 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 633027 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 742701 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 727921 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 874629 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 898183 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 376531 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 640430 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 385486 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 108386 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 771520 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 635816 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 551270 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 181128 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 259378 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 525515 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 148698 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 594781 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 726917 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 733474 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 858624 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 69312 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 369465 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 842635 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 10114 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 676527 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 709892 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 161175 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 235820 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 13873 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 128362 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 656187 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 144333 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 274311 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 703090 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 537267 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 294353 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 608929 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 290900 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 238031 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 686599 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 440641 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 144923 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 151846 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 30485 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 154843 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 99972 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 1549 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 141940 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 116658 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 84820 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 44303 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 78288 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 179292 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 45726 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 28331 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P084513 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P084513 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P084513 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P639460 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P349689 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 18509 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 18135 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 102714 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 14864 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 76891 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 85788 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 28216 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10884988 - -
          Affymetrix Rat Gene 1.0 ST Array 10884990 - -
          Affymetrix Rat Gene 1.0 ST Array 10884991 - -
          Affymetrix Rat Gene 1.0 ST Array 10884992 - -
          Affymetrix Rat Gene 1.0 ST Array 10884982 - -
          Affymetrix Rat Gene 1.0 ST Array 10884989 - -
          Affymetrix Rat Gene 1.0 ST Array 10884987 - -
          Affymetrix Rat Gene 1.0 ST Array 10884983 - -
          Affymetrix Rat Gene 1.0 ST Array 10884986 - -
          Affymetrix Rat Gene 1.0 ST Array 10884993 - -
          Affymetrix Rat Gene 1.0 ST Array 10884985 - -
          Affymetrix Rat Gene 1.0 ST Array 10884980 - -
          Affymetrix Rat Gene 1.0 ST Array 10884984 - -
          Affymetrix Rat Gene 1.0 ST Array 10884981 - -
          Affymetrix Rat Gene 1.0 ST Array 10884979 - -
          Affymetrix Rat Gene 1.1 ST Array 10884978 - -
          Rat Genome 230 2.0 Array 1384007_at - -
          Rat Genome 230 2.0 Array 1397415_PM_at - -
          Rat Genome 230 2.0 Array 1384007_PM_at - -
          Rat Genome 230 2.0 Array 1397415_at - -
          Affymetrix HT Rat Focus Array 1384007_PM_at - -
          Affymetrix HT Rat Focus Array 1397415_at - -
          Affymetrix HT Rat Focus Array 1397415_PM_at - -
          Affymetrix HT Rat Focus Array 1384007_at - -
          Affymetrix Rat Clariom S Assay PSR0600006007.rn.1 TC0600000906.rn.1 RTA TC
          Rat Expression Set 230 1397415_PM_at - -
          Rat Expression Set 230 1397415_at - -
          Rat Expression Set 230 1384007_PM_at - -
          Rat Expression Set 230 1384007_at - -
          Affymetrix Rat Gene 1.0 ST Array 10884978 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21662801 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1376276 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline