Search
Search
| Gene Symbol: Prpf39 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003750171.2 | XP_003750219.1 | 4 - 5 | 721 | 72 | |
| XM_006240140.2 | XP_006240202.1 | 4 - 5 | 1229 | 72 | ||
| XM_006240141.2 | XP_006240203.1 | 5 - 6 | 1126 | 72 | ||
| XM_006240142.2 | XP_006240204.1 | 4 - 5 | 791 | 72 | ||
| XM_006240143.2 | XP_006240205.1 | 4 - 5 | 439 | 72 | ||
| XM_006240144.2 | XP_006240206.1 | 3 - 4 | 375 | 72 | ||
| GenBank mRNA | BC166472.1 | - | 3 - 4 | 1351 | 72 | |
Set Membership: |
|