Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01469119_m1
          See other PRKD1 GE Assays ›
          Gene Symbol:
          Prkd1
          Gene Name:
          protein kinase D1
          Gene Aliases:
          Pkcm, Prkcm, nPKC-D1, nPKC-mu
          Chromosome Location:
          Chr.6: 71035017 - 71349531 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01469119_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Prkd1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001276715.1 NP_001263644.1 18 - 19 2535 69
          XM_006240074.2 XP_006240136.1 17 - 18 2793 69
          GenBank mRNA AB020616.1 - 3 - 4 384 69

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0600002420.rn.1

          Set Membership:

          Amplicon length less than or equal to 70 Apoptosis Protein Kinase 3' Most Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          non-receptor serine/threonine protein kinase

          Gene Ontology Categories:

          Function(s) Process(es)

          angiogenesis
          positive regulation of endothelial cell proliferation
          apoptotic process
          inflammatory response
          Golgi organization
          positive regulation of endothelial cell migration
          regulation of keratinocyte proliferation
          positive regulation of neuron projection development
          peptidyl-serine phosphorylation
          positive regulation of CREB transcription factor activity
          positive regulation of peptidyl-serine phosphorylation
          cellular response to oxidative stress
          intracellular signal transduction
          cellular response to vascular endothelial growth factor stimulus
          positive regulation of endothelial cell chemotaxis by VEGF-activated vascular endothelial growth factor receptor signaling pathway
          positive regulation of I-kappaB kinase/NF-kappaB signaling
          positive regulation of blood vessel endothelial cell migration
          innate immune response
          positive regulation of osteoblast differentiation
          positive regulation of angiogenesis
          positive regulation of transcription from RNA polymerase II promoter
          vascular endothelial growth factor receptor signaling pathway
          Golgi vesicle transport
          positive regulation of NF-kappaB transcription factor activity
          regulation of release of sequestered calcium ion into cytosol
          negative regulation of cell death
          protein kinase D signaling
          positive regulation of histone deacetylase activity
          positive regulation of endothelial cell chemotaxis
          protein serine/threonine kinase activity
          protein kinase C activity
          ATP binding
          identical protein binding
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P124599 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P16038 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P256110 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P463871 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 42900 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 38260 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10889791 - -
          Affymetrix Rat Gene 1.0 ST Array 10889793 - -
          Affymetrix Rat Gene 1.0 ST Array 10889792 - -
          Affymetrix Rat Gene 1.0 ST Array 10889790 - -
          Affymetrix Rat Gene 1.1 ST Array 10889789 - -
          Rat Genome 230 2.0 Array 1381949_at - -
          Rat Genome 230 2.0 Array 1381949_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0600017181.rn.1 TC0600002420.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0600017183.rn.1 TC0600002420.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0600017178.rn.1 TC0600002420.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0600017177.rn.1 TC0600002420.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0600017184.rn.1 TC0600002420.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0600017179.rn.1 TC0600002420.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0600017180.rn.1 TC0600002420.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0600017182.rn.1 TC0600002420.rn.1 RTA TC
          Rat Expression Set 230 1381949_at - -
          Rat Expression Set 230 1381949_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10889789 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21304140 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1373671 - -
          RatRef-12 Expression BeadChip ILMN_48017 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline