Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01469616_m1
          See other PLEKHH1 GE Assays ›
          Gene Symbol:
          Plekhh1
          Gene Name:
          pleckstrin homology domain containing, family H (with MyTH4 domain) member 1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.6: 102215424 - 102264345 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01469616_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Plekhh1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001108036.2 NP_001101506.2 9 - 10 1723 82
          XM_006240262.2 XP_006240324.1 9 - 10 1604 82
          XM_006240263.2 XP_006240325.1 9 - 10 1707 82
          XM_008764775.1 XP_008762997.1 9 - 10 1504 82
          XM_008764776.1 XP_008762998.1 9 - 10 1604 82
          XM_008764777.1 XP_008762999.1 9 - 10 1604 82

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0600001044.rn.1

          Set Membership:

          Amplicon length between 71 and 85 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P108389 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P884935 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 917489 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 602318 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 94591 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 293342 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 505248 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 901510 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 63804 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 74876 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 369027 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 207000 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 781476 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 633719 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 794381 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 856917 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 36734 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 763499 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 827922 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 412329 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 717201 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 185731 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 387778 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 698932 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 277819 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 700725 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 177204 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 118281 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 744850 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 242370 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 717031 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 131448 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 209237 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 489125 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 830017 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 543335 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 312542 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 494707 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 60852 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 709873 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 57319 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 6638 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 147177 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 69077 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 142710 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P108389 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P884935 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P108389 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P884935 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P108389 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P884935 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P492482 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P884935 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 92434 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10885534 - -
          Affymetrix Rat Gene 1.0 ST Array 10885538 - -
          Affymetrix Rat Gene 1.0 ST Array 10885543 - -
          Affymetrix Rat Gene 1.0 ST Array 10885546 - -
          Affymetrix Rat Gene 1.0 ST Array 10885531 - -
          Affymetrix Rat Gene 1.0 ST Array 10885544 - -
          Affymetrix Rat Gene 1.0 ST Array 10885533 - -
          Affymetrix Rat Gene 1.0 ST Array 10885537 - -
          Affymetrix Rat Gene 1.0 ST Array 10885553 - -
          Affymetrix Rat Gene 1.0 ST Array 10885547 - -
          Affymetrix Rat Gene 1.0 ST Array 10885548 - -
          Affymetrix Rat Gene 1.0 ST Array 10885536 - -
          Affymetrix Rat Gene 1.0 ST Array 10885540 - -
          Affymetrix Rat Gene 1.0 ST Array 10885559 - -
          Affymetrix Rat Gene 1.0 ST Array 10885552 - -
          Affymetrix Rat Gene 1.0 ST Array 10885539 - -
          Affymetrix Rat Gene 1.0 ST Array 10885535 - -
          Affymetrix Rat Gene 1.0 ST Array 10885550 - -
          Affymetrix Rat Gene 1.0 ST Array 10885545 - -
          Affymetrix Rat Gene 1.0 ST Array 10885532 - -
          Affymetrix Rat Gene 1.0 ST Array 10885541 - -
          Affymetrix Rat Gene 1.0 ST Array 10885554 - -
          Affymetrix Rat Gene 1.0 ST Array 10885542 - -
          Affymetrix Rat Gene 1.1 ST Array 10885524 - -
          Rat Genome 230 2.0 Array 1384277_at - -
          Rat Genome 230 2.0 Array 1395020_PM_at - -
          Rat Genome 230 2.0 Array 1384277_PM_at - -
          Rat Genome 230 2.0 Array 1395020_at - -
          Affymetrix HT Rat Focus Array 1395020_PM_at - -
          Affymetrix HT Rat Focus Array 1395020_at - -
          Affymetrix Rat Clariom S Assay PSR0600007347.rn.1 TC0600001044.rn.1 RTA TC
          Rat Expression Set 230 1384277_at - -
          Rat Expression Set 230 1395020_PM_at - -
          Rat Expression Set 230 1395020_at - -
          Rat Expression Set 230 1384277_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10885524 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21081788 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1358715 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-wn6hs:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline