Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lbh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001129880.2 | NP_001123352.1 | 2 - 3 | 346 | 114 | |
XM_008764533.1 | XP_008762755.1 | 4 - 5 | 495 | 114 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Rattus norvegicus 1700 Probe spans exons |