Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01471177_m1
          See other SLC22A5 GE Assays ›
          Gene Symbol:
          Slc22a5
          Gene Name:
          solute carrier family 22 (organic cation/carnitine transporter), member 5
          Gene Aliases:
          CT1, OCTN2, UST2r
          Chromosome Location:
          Chr.10: 39201101 - 39228090 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01471177_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Slc22a5
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_019269.1 NP_062142.1 5 - 6 1044 69
          GenBank mRNA AB017260.1 - 5 - 6 1044 69
          AF110416.1 - 5 - 6 1044 69
          AJ001933.1 - 5 - 6 1030 69
          BC166421.1 - 4 - 5 793 9089728 69

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1000002798.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Transporter Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Slc22a5

          Panther Classification:

          Molecular Function -

          secondary carrier transporter

          Gene Ontology Categories:

          Function(s) Process(es)

          amino acid transmembrane transport
          triglyceride metabolic process
          sodium ion transport
          mitochondrion organization
          adult heart development
          locomotory behavior
          carnitine metabolic process
          organic cation transport
          quaternary ammonium group transport
          carnitine transport
          antibiotic transport
          reproductive structure development
          quorum sensing involved in interaction with host
          positive regulation of intestinal epithelial structure maintenance
          sodium-dependent organic cation transport
          modified amino acid transport
          carnitine transmembrane transport
          protein binding
          ATP binding
          carnitine transmembrane transporter activity
          symporter activity
          cation:cation antiporter activity
          quaternary ammonium group transmembrane transporter activity
          PDZ domain binding
          antibiotic transporter activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P1001820 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 943271 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 799384 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 953532 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 47014 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 521950 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 390381 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 732903 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 197574 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 370493 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 145960 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 31583 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 57417 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 348321 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 559083 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 99746 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 945420 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 38612 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 142455 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P1001820 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P1001820 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P1001820 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P1001820 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P473074 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700482091 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 701825421 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 97811 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 15843 - -
          Agilent Rat cDNA Microarray (G4105A) 700482091 - -
          Agilent Rat cDNA Microarray (G4105A) 701825421 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10742650 - -
          Affymetrix Rat Gene 1.0 ST Array 10742656 - -
          Affymetrix Rat Gene 1.0 ST Array 10742648 - -
          Affymetrix Rat Gene 1.0 ST Array 10742653 - -
          Affymetrix Rat Gene 1.0 ST Array 10742654 - -
          Affymetrix Rat Gene 1.0 ST Array 10742655 - -
          Affymetrix Rat Gene 1.0 ST Array 10742652 - -
          Affymetrix Rat Gene 1.0 ST Array 10742649 - -
          Affymetrix Rat Gene 1.0 ST Array 10742647 - -
          Affymetrix Rat Gene 1.0 ST Array 10742646 - -
          Affymetrix Rat Gene 1.0 ST Array 10742651 - -
          Affymetrix Rat Gene 1.1 ST Array 10742645 - -
          Rat Genome 230 2.0 Array 1367950_at - -
          Rat Genome 230 2.0 Array 1391516_at - -
          Rat Genome 230 2.0 Array 1367950_PM_at - -
          Rat Genome 230 2.0 Array 1391516_PM_at - -
          Affymetrix HT Rat Focus Array 1367950_at - -
          Affymetrix HT Rat Focus Array 1367950_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1000024545.rn.1 TC1000002798.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000024544.rn.1 TC1000002798.rn.1 RTA TC
          Rat Expression Set 230 1367950_at - -
          Rat Expression Set 230 1367950_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10742645 - -
          Rat Genome U34 Set AB017260_s_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21858745 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1372579 - -
          RatRef-12 Expression BeadChip ILMN_54600 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-6wvps:80/100.66.75.98:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0