Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01471218_m1
          See other ABCA8A GE Assays ›
          Gene Symbol:
          Abca8a
          Gene Name:
          ATP-binding cassette, subfamily A (ABC1), member 8a
          Gene Aliases:
          -
          Chromosome Location:
          Chr.10: 98319480 - 98390490 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01471218_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Abca8a
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001281824.1 NP_001268753.1 31 - 32 4180 90
          XM_006247611.2 XP_006247673.1 31 - 32 4287 90
          XM_008768370.1 XP_008766592.1 31 - 32 4226 90

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1000004036.rn.1

          Set Membership:

          Amplicon length between 86 and 100 Transporter 3' Most Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          ATP-binding cassette (ABC) transporter primary active transporter transporter

          Gene Ontology Categories:

          Function(s) Process(es)

          transport
          biological_process
          metabolic process
          molecular_function
          transporter activity
          ATP binding
          ATPase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P594228 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P594228 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P594228 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P594228 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P17127 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10748633 - -
          Affymetrix Rat Gene 1.0 ST Array 10748605 - -
          Affymetrix Rat Gene 1.0 ST Array 10748617 - -
          Affymetrix Rat Gene 1.0 ST Array 10748628 - -
          Affymetrix Rat Gene 1.0 ST Array 10748631 - -
          Affymetrix Rat Gene 1.0 ST Array 10748625 - -
          Affymetrix Rat Gene 1.0 ST Array 10748638 - -
          Affymetrix Rat Gene 1.0 ST Array 10748634 - -
          Affymetrix Rat Gene 1.0 ST Array 10748612 - -
          Affymetrix Rat Gene 1.0 ST Array 10748616 - -
          Affymetrix Rat Gene 1.0 ST Array 10748604 - -
          Affymetrix Rat Gene 1.0 ST Array 10748635 - -
          Affymetrix Rat Gene 1.0 ST Array 10748606 - -
          Affymetrix Rat Gene 1.0 ST Array 10748603 - -
          Affymetrix Rat Gene 1.0 ST Array 10748607 - -
          Affymetrix Rat Gene 1.0 ST Array 10748609 - -
          Affymetrix Rat Gene 1.0 ST Array 10748608 - -
          Affymetrix Rat Gene 1.0 ST Array 10748622 - -
          Affymetrix Rat Gene 1.0 ST Array 10748623 - -
          Affymetrix Rat Gene 1.0 ST Array 10748611 - -
          Affymetrix Rat Gene 1.0 ST Array 10748620 - -
          Affymetrix Rat Gene 1.0 ST Array 10748619 - -
          Affymetrix Rat Gene 1.0 ST Array 10748610 - -
          Affymetrix Rat Gene 1.0 ST Array 10748615 - -
          Affymetrix Rat Gene 1.0 ST Array 10748614 - -
          Affymetrix Rat Gene 1.0 ST Array 10748618 - -
          Affymetrix Rat Gene 1.0 ST Array 10748637 - -
          Affymetrix Rat Gene 1.0 ST Array 10748602 - -
          Affymetrix Rat Gene 1.0 ST Array 10748636 - -
          Affymetrix Rat Gene 1.0 ST Array 10748621 - -
          Affymetrix Rat Gene 1.0 ST Array 10748632 - -
          Affymetrix Rat Gene 1.0 ST Array 10748624 - -
          Affymetrix Rat Gene 1.0 ST Array 10748613 - -
          Affymetrix Rat Gene 1.0 ST Array 10748626 - -
          Affymetrix Rat Gene 1.0 ST Array 10748630 - -
          Affymetrix Rat Gene 1.0 ST Array 10748627 - -
          Rat Genome 230 2.0 Array 1390783_PM_at - -
          Rat Genome 230 2.0 Array 1390783_at - -
          Affymetrix HT Rat Focus Array 1390783_at - -
          Affymetrix HT Rat Focus Array 1390783_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1000036609.rn.1 TC1000004036.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000036611.rn.1 TC1000004036.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000036606.rn.1 TC1000004036.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000036610.rn.1 TC1000004036.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000036612.rn.1 TC1000004036.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000036608.rn.1 TC1000004036.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000036607.rn.1 TC1000004036.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000036613.rn.1 TC1000004036.rn.1 RTA TC
          Rat Expression Set 230 1390783_at - -
          Rat Expression Set 230 1390783_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10748601 - -
          Rat Genome U34 Set rc_AI009456_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21452990 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1367219 - -
          RatRef-12 Expression BeadChip ILMN_55476 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline