Search
Search

Slc25a39| Gene Symbol: Slc25a39 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024792.1 | NP_001019963.1 | 2 - 3 | 196 | 84 | |
| XM_006247374.2 | XP_006247436.1 | 2 - 3 | 534 | 84 | ||
| XM_006247376.2 | XP_006247438.1 | 2 - 3 | 219 | 84 | ||
| GenBank mRNA | BC081862.1 | - | 2 - 3 | 222 | 84 | |
| BC097349.1 | - | 2 - 3 | 196 | 7460172 | 84 | |
Set Membership: |
|