Search
Search
| Gene Symbol: Elp5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001001718.1 | NP_001001718.1 | 5 - 6 | 849 | 63 | |
| XM_006246639.2 | XP_006246701.1 | 6 - 7 | 1049 | 63 | ||
| XM_006246640.1 | XP_006246702.1 | 4 - 5 | 358 | 63 | ||
| GenBank mRNA | AY623900.1 | - | 5 - 6 | 849 | 63 | |
| BC158569.1 | - | 6 - 7 | 1052 | 9101477 | 63 | |
Set Membership: |
|