Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01471640_g1
          See other ATPAF2 GE Assays ›
          Gene Symbol:
          Atpaf2
          Gene Name:
          ATP synthase mitochondrial F1 complex assembly factor 2
          Gene Aliases:
          -
          Chromosome Location:
          Chr.10: 46768539 - 46783889 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01471640_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Atpaf2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001107006.1 NP_001100476.1 6 - 7 774 63

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1000002934.rn.1

          Set Membership:

          Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          chaperone

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          proton-transporting ATP synthase complex assembly
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P140093 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 787188 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 810961 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 30890 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 573380 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 572427 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 109011 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 598875 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 187432 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 628160 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 56442 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 43657 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P140093 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P140093 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P140093 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P218247 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 5963 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10743262 - -
          Affymetrix Rat Gene 1.0 ST Array 10743259 - -
          Affymetrix Rat Gene 1.0 ST Array 10743255 - -
          Affymetrix Rat Gene 1.0 ST Array 10743261 - -
          Affymetrix Rat Gene 1.0 ST Array 10743258 - -
          Affymetrix Rat Gene 1.0 ST Array 10743256 - -
          Affymetrix Rat Gene 1.0 ST Array 10743260 - -
          Affymetrix Rat Gene 1.0 ST Array 10743257 - -
          Affymetrix Rat Gene 1.1 ST Array 10743254 - -
          Rat Genome 230 2.0 Array 1371892_PM_at - -
          Rat Genome 230 2.0 Array 1371892_at - -
          Affymetrix HT Rat Focus Array 1371892_at - -
          Affymetrix HT Rat Focus Array 1371892_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1000025774.rn.1 TC1000002934.rn.1 RTA TC
          Rat Expression Set 230 1371892_PM_at - -
          Rat Expression Set 230 1371892_at - -
          Affymetrix Rat Gene 1.0 ST Array 10743254 - -
          Rat Genome U34 Set rc_AI177136_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21604931 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1358633 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-4lm45:80/100.66.75.98:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0