Search
Search

Ccdc103| Gene Symbol: Ccdc103 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109049.1 | NP_001102519.1 | 4 - 5 | 466 | 87 | |
| XM_006247538.2 | XP_006247600.1 | 3 - 4 | 320 | 87 | ||
| XM_006247540.2 | XP_006247602.1 | 2 - 3 | 291 | 87 | ||
Set Membership: |
|