Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01471788_m1
          See other SHPK GE Assays ›
          Gene Symbol:
          Shpk
          Gene Name:
          sedoheptulokinase
          Gene Aliases:
          Carkl
          Chromosome Location:
          Chr.10: 59765328 - 59789676 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01471788_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Shpk
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001033682.1 NP_001028854.1 5 - 6 852 77
          GenBank mRNA BC101914.1 - 5 - 6 852 7935768 77

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1000001024.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Shpk

          Panther Classification:

          Molecular Function -

          carbohydrate kinase

          Gene Ontology Categories:

          Function(s) Process(es)

          carbohydrate metabolic process
          pentose-phosphate shunt, non-oxidative branch
          phosphorylation
          cellular response to interleukin-13
          regulation of macrophage activation
          regulation of inflammatory response
          cellular response to lipopolysaccharide
          cellular response to interleukin-4
          sedoheptulokinase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P663350 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 295712 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 262428 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 278883 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 216957 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 112213 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 436772 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 104208 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 777797 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 697810 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 218529 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 898635 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 394730 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 111636 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 630896 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 783299 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 625632 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 684456 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 685738 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 428174 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 109820 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 12669 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 52503 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 108601 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 39221 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P663350 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P663350 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P663350 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P19057 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 51709 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 83690 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 19866 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10735675 - -
          Affymetrix Rat Gene 1.0 ST Array 10735674 - -
          Affymetrix Rat Gene 1.0 ST Array 10735672 - -
          Affymetrix Rat Gene 1.0 ST Array 10735671 - -
          Affymetrix Rat Gene 1.0 ST Array 10735670 - -
          Affymetrix Rat Gene 1.0 ST Array 10735673 - -
          Affymetrix Rat Gene 1.0 ST Array 10735676 - -
          Affymetrix Rat Gene 1.1 ST Array 10735669 - -
          Rat Genome 230 2.0 Array 1384273_PM_at - -
          Rat Genome 230 2.0 Array 1384273_at - -
          Affymetrix HT Rat Focus Array 1384273_at - -
          Affymetrix HT Rat Focus Array 1384273_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1000009364.rn.1 TC1000001024.rn.1 RTA TC
          Rat Expression Set 230 1384273_at - -
          Rat Expression Set 230 1384273_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10735669 - -
          Rat Genome U34 Set rc_AI070132_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21343848 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1366109 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline