Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01471850_g1
          See other ALDOC GE Assays ›
          Gene Symbol:
          Aldoc
          Gene Name:
          aldolase C, fructose-bisphosphate
          Gene Aliases:
          ALDCAA, F16dip7, RATALDCAA
          Chromosome Location:
          Chr.10: 65586504 - 65590093 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01471850_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Aldoc
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_012497.1 NP_036629.1 8 - 9 1106 81
          GenBank mRNA BC059154.1 - 5 - 6 568 81
          BC099749.1 - 8 - 9 1092 5599032 81
          FQ212739.1 - 2 - 3 373 81
          FQ213260.1 - 2 - 3 373 81
          FQ213506.1 - 8 - 9 1109 81
          FQ214420.1 - 8 - 9 1132 81
          FQ228893.1 - 8 - 9 1106 81

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1000004359.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Aldoc

          Panther Classification:

          Molecular Function -

          aldolase

          Gene Ontology Categories:

          Function(s) Process(es)

          response to hypoxia
          glycolytic process
          apoptotic process
          aging
          response to organonitrogen compound
          response to organic cyclic compound
          fructose 1,6-bisphosphate metabolic process
          epithelial cell differentiation
          organ regeneration
          protein homotetramerization
          protein heterotetramerization
          fructose-bisphosphate aldolase activity
          protein binding
          cytoskeletal protein binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P056558 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P064909 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 434961 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 506699 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 145371 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P056558 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P064909 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P056558 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P064909 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P064909 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P056558 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P512996 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700244954 - -
          Agilent Rat cDNA Microarray (G4105A) 700244954 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10745103 - -
          Affymetrix Rat Gene 1.0 ST Array 10745098 - -
          Affymetrix Rat Gene 1.0 ST Array 10745104 - -
          Affymetrix Rat Gene 1.0 ST Array 10745099 - -
          Affymetrix Rat Gene 1.0 ST Array 10745101 - -
          Affymetrix Rat Gene 1.0 ST Array 10745106 - -
          Affymetrix Rat Gene 1.0 ST Array 10745105 - -
          Affymetrix Rat Gene 1.0 ST Array 10745100 - -
          Affymetrix Rat Gene 1.0 ST Array 10745096 - -
          Affymetrix Rat Gene 1.0 ST Array 10745097 - -
          Affymetrix Rat Gene 1.0 ST Array 10745102 - -
          Affymetrix Rat Gene 1.1 ST Array 10745095 - -
          Rat Genome 230 2.0 Array 1386998_at - -
          Rat Genome 230 2.0 Array 1386998_PM_at - -
          Affymetrix HT Rat Focus Array 1386998_PM_at - -
          Affymetrix HT Rat Focus Array 1386998_at - -
          Affymetrix Rat Clariom S Assay PSR1000040456.rn.1 TC1000004359.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000040457.rn.1 TC1000004359.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000040459.rn.1 TC1000004359.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000040458.rn.1 TC1000004359.rn.1 RTA TC
          Rat Expression Set 230 1386998_PM_at - -
          Rat Expression Set 230 1386998_at - -
          Affymetrix Rat Gene 1.0 ST Array 10745095 - -
          Rat Genome U34 Set M63656_s_at - -
          Rat Genome U34 Set X06984cds_s_at - -
          Rat Genome U34 Set rc_AI180121_s_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21801520 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1349452 - -
          RatRef-12 Expression BeadChip ILMN_53421 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline