Search
Search

RGD1564036| Gene Symbol: RGD1564036 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109030.2 | NP_001102500.2 | 2 - 3 | 131 | 115 | |
| XM_008767694.1 | XP_008765916.1 | 2 - 3 | 705 | 115 | ||
| GenBank mRNA | BC166832.1 | - | 2 - 3 | 131 | 8375915 | 115 |
Set Membership: |
|