Search
Search
| Gene Symbol: Btk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007798.1 | NP_001007799.1 | 7 - 8 | 842 | 81 | |
| GenBank mRNA | AY787790.1 | - | 7 - 8 | 842 | 81 | |
| FQ215667.1 | - | 986 | 81 | |||
Set Membership: |
|