Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01472419_m1
          See other DDX1 GE Assays ›
          Gene Symbol:
          Ddx1
          Gene Name:
          DEAD (Asp-Glu-Ala-Asp) box helicase 1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.6: 38422892 - 38453762 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01472419_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ddx1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_053414.1 NP_445866.1 23 - 24 1921 107
          GenBank mRNA BC082049.1 - 23 - 24 1921 7131804 107
          FQ226373.1 - 11 - 12 946 107

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0600002151.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Rattus norvegicus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ddx1

          Gene Ontology Categories:

          Function(s) Process(es)

          double-strand break repair
          transcription, DNA-templated
          regulation of transcription, DNA-templated
          tRNA splicing, via endonucleolytic cleavage and ligation
          mRNA processing
          multicellular organism development
          response to virus
          DNA duplex unwinding
          response to exogenous dsRNA
          nucleic acid phosphodiester bond hydrolysis
          DNA binding
          chromatin binding
          transcription cofactor activity
          double-stranded RNA binding
          nuclease activity
          exonuclease activity
          ATP binding
          ATP-dependent helicase activity
          poly(A) binding
          DNA/RNA helicase activity
          poly(A) RNA binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P456852 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 172829 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 835178 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 302885 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 916649 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 528542 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 478965 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 498999 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 883965 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 52536 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 241213 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 501271 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 772082 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 741715 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 278000 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 452551 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 269473 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 95296 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 628693 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 534975 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 621270 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 843171 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 748277 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 272661 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 837752 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 86434 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 128540 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 5228 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 173645 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 180583 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 90607 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 5765 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 133193 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P456852 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P456852 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P456852 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P456852 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P15623 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P231497 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 60710 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 2055 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 102566 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 3520 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 58075 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10889252 - -
          Affymetrix Rat Gene 1.0 ST Array 10889255 - -
          Affymetrix Rat Gene 1.0 ST Array 10889240 - -
          Affymetrix Rat Gene 1.0 ST Array 10889237 - -
          Affymetrix Rat Gene 1.0 ST Array 10889254 - -
          Affymetrix Rat Gene 1.0 ST Array 10889239 - -
          Affymetrix Rat Gene 1.0 ST Array 10889251 - -
          Affymetrix Rat Gene 1.0 ST Array 10889238 - -
          Affymetrix Rat Gene 1.0 ST Array 10889253 - -
          Affymetrix Rat Gene 1.0 ST Array 10889259 - -
          Affymetrix Rat Gene 1.0 ST Array 10889246 - -
          Affymetrix Rat Gene 1.0 ST Array 10889250 - -
          Affymetrix Rat Gene 1.0 ST Array 10889256 - -
          Affymetrix Rat Gene 1.0 ST Array 10889241 - -
          Affymetrix Rat Gene 1.0 ST Array 10889257 - -
          Affymetrix Rat Gene 1.0 ST Array 10889242 - -
          Affymetrix Rat Gene 1.0 ST Array 10889249 - -
          Affymetrix Rat Gene 1.0 ST Array 10889248 - -
          Affymetrix Rat Gene 1.0 ST Array 10889236 - -
          Affymetrix Rat Gene 1.0 ST Array 10889244 - -
          Affymetrix Rat Gene 1.0 ST Array 10889235 - -
          Affymetrix Rat Gene 1.0 ST Array 10889245 - -
          Affymetrix Rat Gene 1.0 ST Array 10889243 - -
          Affymetrix Rat Gene 1.0 ST Array 10889258 - -
          Affymetrix Rat Gene 1.0 ST Array 10889247 - -
          Affymetrix Rat Gene 1.1 ST Array 10889234 - -
          Rat Genome 230 2.0 Array 1373319_PM_at - -
          Rat Genome 230 2.0 Array 1373319_at - -
          Affymetrix HT Rat Focus Array 1373319_PM_at - -
          Affymetrix HT Rat Focus Array 1373319_at - -
          Affymetrix Rat Clariom S Assay PSR0600015589.rn.1 TC0600002151.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0600015591.rn.1 TC0600002151.rn.1 RTA TC
          Rat Expression Set 230 1373319_PM_at - -
          Rat Expression Set 230 1373319_at - -
          Affymetrix Rat Gene 1.0 ST Array 10889234 - -
          Rat Genome U34 Set rc_AI172325_at - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1351457 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline