Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01472509_m1
          See other RALGAPA1 GE Assays ›
          Gene Symbol:
          Ralgapa1
          Gene Name:
          Ral GTPase activating protein, alpha subunit 1 (catalytic)
          Gene Aliases:
          GRIPE, Garnl1, Tulip1, p240
          Chromosome Location:
          Chr.6: 76386975 - 76661529 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01472509_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ralgapa1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AF041106.1 - 24 - 25 3594 85
          AF041107.1 - 16 - 17 2720 85
          FQ214235.1 - 3 - 4 404 85

          Back To Top

          More Information


          Set Membership:

          Amplicon length between 71 and 85 3' Most Recommended Assay Rattus norvegicus Probe spans exons

          Panther Classification:

          Molecular Function -

          GTPase-activating protein

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P120709 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P539680 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 47231 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P120709 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P539680 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P120709 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P539680 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P120709 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P539680 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P153026 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P539680 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P276475 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P276477 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P440466 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 702170548 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700695931 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 4622 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 21300 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 45886 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 24567 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 31406 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 10044 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 13834 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 76320 - -
          Agilent Rat cDNA Microarray (G4105A) 700695931 - -
          Agilent Rat cDNA Microarray (G4105A) 702170548 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10890051 - -
          Affymetrix Rat Gene 1.0 ST Array 10890057 - -
          Affymetrix Rat Gene 1.0 ST Array 10890058 - -
          Affymetrix Rat Gene 1.0 ST Array 10890052 - -
          Affymetrix Rat Gene 1.0 ST Array 10890047 - -
          Affymetrix Rat Gene 1.0 ST Array 10890049 - -
          Affymetrix Rat Gene 1.0 ST Array 10890033 - -
          Affymetrix Rat Gene 1.0 ST Array 10890048 - -
          Affymetrix Rat Gene 1.0 ST Array 10890055 - -
          Affymetrix Rat Gene 1.0 ST Array 10890040 - -
          Affymetrix Rat Gene 1.0 ST Array 10890070 - -
          Affymetrix Rat Gene 1.0 ST Array 10890067 - -
          Affymetrix Rat Gene 1.0 ST Array 10890054 - -
          Affymetrix Rat Gene 1.0 ST Array 10890038 - -
          Affymetrix Rat Gene 1.0 ST Array 10890032 - -
          Affymetrix Rat Gene 1.0 ST Array 10890063 - -
          Affymetrix Rat Gene 1.0 ST Array 10890066 - -
          Affymetrix Rat Gene 1.0 ST Array 10890059 - -
          Affymetrix Rat Gene 1.0 ST Array 10890060 - -
          Affymetrix Rat Gene 1.0 ST Array 10890053 - -
          Affymetrix Rat Gene 1.0 ST Array 10890042 - -
          Affymetrix Rat Gene 1.0 ST Array 10890035 - -
          Affymetrix Rat Gene 1.0 ST Array 10890039 - -
          Affymetrix Rat Gene 1.0 ST Array 10890046 - -
          Affymetrix Rat Gene 1.0 ST Array 10890050 - -
          Affymetrix Rat Gene 1.0 ST Array 10890064 - -
          Affymetrix Rat Gene 1.0 ST Array 10890061 - -
          Affymetrix Rat Gene 1.0 ST Array 10890036 - -
          Affymetrix Rat Gene 1.0 ST Array 10890069 - -
          Affymetrix Rat Gene 1.0 ST Array 10890062 - -
          Affymetrix Rat Gene 1.0 ST Array 10890068 - -
          Affymetrix Rat Gene 1.0 ST Array 10890065 - -
          Affymetrix Rat Gene 1.0 ST Array 10890045 - -
          Affymetrix Rat Gene 1.0 ST Array 10890041 - -
          Affymetrix Rat Gene 1.0 ST Array 10890056 - -
          Affymetrix Rat Gene 1.0 ST Array 10890037 - -
          Affymetrix Rat Gene 1.0 ST Array 10890044 - -
          Affymetrix Rat Gene 1.0 ST Array 10890072 - -
          Affymetrix Rat Gene 1.0 ST Array 10890043 - -
          Affymetrix Rat Gene 1.0 ST Array 10890034 - -
          Affymetrix Rat Gene 1.1 ST Array 10890031 - -
          Affymetrix Rat Gene 1.1 ST Array 10884669 - -
          Rat Genome 230 2.0 Array 1376338_at - -
          Rat Genome 230 2.0 Array 1390217_at - -
          Rat Genome 230 2.0 Array 1395385_at - -
          Rat Genome 230 2.0 Array 1368995_PM_at - -
          Rat Genome 230 2.0 Array 1387405_PM_at - -
          Rat Genome 230 2.0 Array 1376338_PM_at - -
          Rat Genome 230 2.0 Array 1368995_at - -
          Rat Genome 230 2.0 Array 1395385_PM_at - -
          Rat Genome 230 2.0 Array 1375670_PM_at - -
          Rat Genome 230 2.0 Array 1368994_a_at - -
          Rat Genome 230 2.0 Array 1387405_at - -
          Rat Genome 230 2.0 Array 1375670_at - -
          Rat Genome 230 2.0 Array 1390217_PM_at - -
          Rat Genome 230 2.0 Array 1368994_PM_a_at - -
          Affymetrix HT Rat Focus Array 1395385_at - -
          Affymetrix HT Rat Focus Array 1368994_PM_a_at - -
          Affymetrix HT Rat Focus Array 1368995_at - -
          Affymetrix HT Rat Focus Array 1368995_PM_at - -
          Affymetrix HT Rat Focus Array 1390217_at - -
          Affymetrix HT Rat Focus Array 1387405_PM_at - -
          Affymetrix HT Rat Focus Array 1390217_PM_at - -
          Affymetrix HT Rat Focus Array 1368994_a_at - -
          Affymetrix HT Rat Focus Array 1395385_PM_at - -
          Affymetrix HT Rat Focus Array 1387405_at - -
          Rat Expression Set 230 1376338_at - -
          Rat Expression Set 230 1390217_PM_at - -
          Rat Expression Set 230 1375670_PM_at - -
          Rat Expression Set 230 1368995_PM_at - -
          Rat Expression Set 230 1387405_at - -
          Rat Expression Set 230 1368995_at - -
          Rat Expression Set 230 1387405_PM_at - -
          Rat Expression Set 230 1368994_a_at - -
          Rat Expression Set 230 1375670_at - -
          Rat Expression Set 230 1368994_PM_a_at - -
          Rat Expression Set 230 1390217_at - -
          Rat Expression Set 230 1376338_PM_at - -
          Rat Expression Set 230 1395385_at - -
          Rat Expression Set 230 1395385_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10890031 - -
          Rat Genome U34 Set AF041106_at - -
          Rat Genome U34 Set AF041107_at - -
          Rat Genome U34 Set AF041107_g_at - -
          Rat Genome U34 Set rc_AI231807_g_at - -
          Rat Genome U34 Set rc_AI233383_at - -
          Rat Genome U34 Set rc_AI170342_at - -
          Rat Genome U34 Set rc_AI100780_at - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1368996 - -
          RatRef-12 Expression BeadChip ILMN_57848 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-6wvps:80/100.66.75.98:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0