Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01473056_m1
          See other TMEM178A GE Assays ›
          Gene Symbol:
          Tmem178a
          Gene Name:
          transmembrane protein 178A
          Gene Aliases:
          Tmem178
          Chromosome Location:
          Chr.6: 3657355 - 3716079 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01473056_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tmem178a
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001004282.1 NP_001004282.1 3 - 4 662 61
          GenBank mRNA BC079338.1 - 3 - 4 692 7106500 61

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0600000052.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Tmem178a

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P130583 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 197948 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 364089 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 249012 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 772331 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 872577 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 676161 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 766906 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 23502 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 968842 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 259368 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 667026 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 674963 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 687744 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 719065 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 226474 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 200775 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 94459 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 125761 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 767507 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 228826 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 463828 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 910718 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 289586 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 57989 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 621290 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 472340 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 599407 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 374097 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 40222 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 722387 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 888292 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 338435 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 101224 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 183646 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 252305 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 595496 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 195551 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 829713 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 324015 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 751646 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 56415 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 291770 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 88298 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 58633 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 20025 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 71046 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 38622 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 127582 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P130583 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P130583 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P130583 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P555072 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 51621 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 35532 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 74095 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 49314 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 10375 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10882527 - -
          Affymetrix Rat Gene 1.0 ST Array 10882529 - -
          Affymetrix Rat Gene 1.0 ST Array 10882528 - -
          Affymetrix Rat Gene 1.0 ST Array 10882526 - -
          Affymetrix Rat Gene 1.1 ST Array 10882525 - -
          Rat Genome 230 2.0 Array 1376106_at - -
          Rat Genome 230 2.0 Array 1376106_PM_at - -
          Affymetrix HT Rat Focus Array 1376106_at - -
          Affymetrix HT Rat Focus Array 1376106_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0600000359.rn.1 TC0600000052.rn.1 RTA TC
          Rat Expression Set 230 1376106_PM_at - -
          Rat Expression Set 230 1376106_at - -
          Affymetrix Rat Gene 1.0 ST Array 10882525 - -
          Rat Genome U34 Set rc_AI010157_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21261216 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1348808 - -
          RatRef-12 Expression BeadChip ILMN_52417 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline