Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01473128_m1
          See other DUS4L GE Assays ›
          Gene Symbol:
          Dus4l
          Gene Name:
          dihydrouridine synthase 4-like (S. cerevisiae)
          Gene Aliases:
          RGD1311445
          Chromosome Location:
          Chr.6: 50929156 - 50943488 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01473128_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Dus4l
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001135803.1 NP_001129275.1 6 - 7 882 68
          XM_006239990.2 XP_006240052.1 6 - 7 813 68
          XM_006239991.1 XP_006240053.1 7 - 8 1040 68
          XM_006239992.2 XP_006240054.1 7 - 8 1317 68
          XM_006239993.2 XP_006240055.1 5 - 6 726 68
          XM_008764601.1 XP_008762823.1 5 - 6 688 68

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0600002279.rn.1

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Recommended Assay Rattus norvegicus 1700 Probe spans exons

          Panther Classification:

          Molecular Function -

          RNA processing factor

          Gene Ontology Categories:

          Function(s) Process(es)

          tRNA dihydrouridine synthesis
          oxidation-reduction process
          tRNA dihydrouridine synthase activity
          flavin adenine dinucleotide binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P157385 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 418316 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 663055 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 262054 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 76133 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 442432 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 161479 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 811264 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 861761 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 650541 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 342107 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 90505 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P157385 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P157385 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P157385 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P363757 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10889572 - -
          Affymetrix Rat Gene 1.0 ST Array 10889570 - -
          Affymetrix Rat Gene 1.0 ST Array 10889569 - -
          Affymetrix Rat Gene 1.0 ST Array 10889573 - -
          Affymetrix Rat Gene 1.0 ST Array 10889571 - -
          Affymetrix Rat Gene 1.1 ST Array 10889568 - -
          Rat Genome 230 2.0 Array 1379551_at - -
          Rat Genome 230 2.0 Array 1379551_PM_at - -
          Rat Genome 230 2.0 Array 1385332_PM_at - -
          Rat Genome 230 2.0 Array 1385332_at - -
          Affymetrix HT Rat Focus Array 1385332_PM_at - -
          Affymetrix HT Rat Focus Array 1379551_PM_at - -
          Affymetrix HT Rat Focus Array 1379551_at - -
          Affymetrix HT Rat Focus Array 1385332_at - -
          Affymetrix Rat Clariom S Assay PSR0600016495.rn.1 TC0600002279.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0600016496.rn.1 TC0600002279.rn.1 RTA TC
          Rat Expression Set 230 1385332_at - -
          Rat Expression Set 230 1379551_at - -
          Rat Expression Set 230 1385332_PM_at - -
          Rat Expression Set 230 1379551_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10889568 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21681661 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1651062 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline