Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • Enterprise Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01473413_m1
          See other DYNC2LI1 GE Assays ›
          Gene Symbol:
          Dync2li1
          Gene Name:
          dynein cytoplasmic 2 light intermediate chain 1
          Gene Aliases:
          RGD1310286
          Chromosome Location:
          Chr.6: 7900963 - 7933795 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01473413_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Dync2li1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001013940.1 NP_001013962.1 11 - 12 1370 77
          XM_006239695.2 XP_006239757.1 12 - 13 1130 77
          GenBank mRNA BC079201.1 - 11 - 12 1370 7131728 77

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0600000102.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Dync2li1

          Panther Classification:

          Molecular Function -

          cytoskeletal protein microtubule or microtubule-binding cytoskeletal protein

          Gene Ontology Categories:

          Function(s) Process(es)

          determination of left/right symmetry
          metabolic process
          cilium assembly
          motor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P23209 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 420510 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 72041 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 698792 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 15699 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 439401 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 631664 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 435979 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 388590 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 490947 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 49936 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 217820 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 123182 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 480170 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 608988 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 943624 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 60729 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 608784 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 70724 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 116584 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 505022 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 410748 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 289033 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 42375 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 107854 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P23209 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P23209 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P23209 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P23209 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 50887 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 94462 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 66046 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 37840 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10882641 - -
          Affymetrix Rat Gene 1.0 ST Array 10882631 - -
          Affymetrix Rat Gene 1.0 ST Array 10882635 - -
          Affymetrix Rat Gene 1.0 ST Array 10882636 - -
          Affymetrix Rat Gene 1.0 ST Array 10882633 - -
          Affymetrix Rat Gene 1.0 ST Array 10882630 - -
          Affymetrix Rat Gene 1.0 ST Array 10882632 - -
          Affymetrix Rat Gene 1.0 ST Array 10882642 - -
          Affymetrix Rat Gene 1.0 ST Array 10882628 - -
          Affymetrix Rat Gene 1.0 ST Array 10882638 - -
          Affymetrix Rat Gene 1.0 ST Array 10882639 - -
          Affymetrix Rat Gene 1.0 ST Array 10882637 - -
          Affymetrix Rat Gene 1.0 ST Array 10882634 - -
          Affymetrix Rat Gene 1.0 ST Array 10882640 - -
          Affymetrix Rat Gene 1.0 ST Array 10882629 - -
          Affymetrix Rat Gene 1.1 ST Array 10882627 - -
          Rat Genome 230 2.0 Array 1374573_at - -
          Rat Genome 230 2.0 Array 1374573_PM_at - -
          Affymetrix HT Rat Focus Array 1374573_PM_at - -
          Affymetrix HT Rat Focus Array 1374573_at - -
          Affymetrix Rat Clariom S Assay PSR0600000595.rn.1 TC0600000102.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0600000596.rn.1 TC0600000102.rn.1 RTA TC
          Rat Expression Set 230 1374573_at - -
          Rat Expression Set 230 1374573_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10882627 - -
          Rat Genome U34 Set rc_AA996695_i_at - -
          Rat Genome U34 Set rc_AA996695_f_at - -
          Rat Genome U34 Set rc_AI177135_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22361918 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1352029 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ckh64:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0