Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01473684_m1
          See other RMDN2 GE Assays ›
          Gene Symbol:
          Rmdn2
          Gene Name:
          regulator of microtubule dynamics 2
          Gene Aliases:
          Fam82a, Fam82a1
          Chromosome Location:
          Chr.6: 2215059 - 2292289 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01473684_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rmdn2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001037200.1 NP_001032277.1 5 - 6 1181 139
          XM_006239633.2 XP_006239695.1 6 - 7 3526 139
          XM_006239634.2 XP_006239696.1 6 - 7 3526 139
          XM_006239635.2 XP_006239697.1 7 - 8 1645 139
          XM_008764422.1 XP_008762644.1 6 - 7 3526 139
          XM_008764423.1 XP_008762645.1 7 - 8 1663 139
          GenBank mRNA BC100261.1 - 5 - 6 1181 7368645 139

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0600000022.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Rmdn2

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P061820 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 638783 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 199401 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 902783 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 496629 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 802735 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 445579 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 269188 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 338257 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 935952 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 942054 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 833623 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 460741 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 265125 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 557866 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 761995 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 139322 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 87826 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 308417 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 504671 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 675272 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 712587 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 715848 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 7432 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 524399 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 270822 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 111356 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 337053 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 748347 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 932577 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 815859 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 850939 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 113835 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 504174 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 4537 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 634524 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 866479 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 700854 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 210671 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 766454 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 668338 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 760703 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 588509 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 145655 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 700370 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 552831 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 121829 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 73400 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 121885 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 180651 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 162283 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 162614 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P061820 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P061820 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P061820 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P506538 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 42990 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 12500 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 12129 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 39651 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 45990 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10882486 - -
          Affymetrix Rat Gene 1.0 ST Array 10882483 - -
          Affymetrix Rat Gene 1.0 ST Array 10882484 - -
          Affymetrix Rat Gene 1.0 ST Array 10882489 - -
          Affymetrix Rat Gene 1.0 ST Array 10882487 - -
          Affymetrix Rat Gene 1.0 ST Array 10882485 - -
          Affymetrix Rat Gene 1.0 ST Array 10882477 - -
          Affymetrix Rat Gene 1.0 ST Array 10882481 - -
          Affymetrix Rat Gene 1.0 ST Array 10882476 - -
          Affymetrix Rat Gene 1.0 ST Array 10882482 - -
          Affymetrix Rat Gene 1.0 ST Array 10882488 - -
          Affymetrix Rat Gene 1.1 ST Array 10882475 - -
          Rat Genome 230 2.0 Array 1379636_PM_at - -
          Rat Genome 230 2.0 Array 1395983_PM_at - -
          Rat Genome 230 2.0 Array 1395983_at - -
          Rat Genome 230 2.0 Array 1379636_at - -
          Affymetrix HT Rat Focus Array 1379636_PM_at - -
          Affymetrix HT Rat Focus Array 1395983_PM_at - -
          Affymetrix HT Rat Focus Array 1395983_at - -
          Affymetrix HT Rat Focus Array 1379636_at - -
          Affymetrix Rat Clariom S Assay PSR0600000184.rn.1 TC0600000022.rn.1 RTA TC
          Rat Expression Set 230 1379636_PM_at - -
          Rat Expression Set 230 1379636_at - -
          Rat Expression Set 230 1395983_PM_at - -
          Rat Expression Set 230 1395983_at - -
          Affymetrix Rat Gene 1.0 ST Array 10882475 - -
          Rat Genome U34 Set rc_AI235895_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21660864 - -
          ABI Rat Genome Survey Microarray 21755468 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1375002 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-hznln:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline