Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture and Transfection
    • Chemicals
    • Chromatography
    • Electron Microscopes
    • Lab Plasticware and Supplies
    • Lab Centrifuges
    • Lab Solutions
    • Mass Spectrometers
    • Next Generation Sequencers
    • See all product categories
  • Applications
    • Brands
    • Industrial and Applied Sciences
    • Food and Beverage
    • Forensics
    • Lab Solutions
    • Life Sciences
    • Pharma and Biopharma
    • Biotechnology
    • Clinical and Diagnostics
    • Digital Solutions
    • See all applications
  • Services
    • Lab Instrument and Equipment Services
    • Custom Services
    • Training Services
    • Financial and Leasing Services
    • Enterprise Level Lab Informatics
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Cell Biology Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • Contact Us
    • Product Documentation
    • Knowledge Base and Product FAQs
    • Learning Centers
    • Supply Center
    • eProcurement Solutions
    • Lab Instrument and Equipment Support
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01473803_m1
          See other TRPA1 GE Assays ›
          Gene Symbol:
          Trpa1
          Gene Name:
          transient receptor potential cation channel, subfamily A, member 1
          Gene Aliases:
          Anktm1
          Chromosome Location:
          Chr.5: 3783247 - 3836485 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01473803_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Trpa1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_207608.1 NP_997491.1 2 - 3 273 104
          GenBank mRNA AY496961.1 - 2 - 3 273 104

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0500000036.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Trpa1

          Panther Classification:

          Molecular Function -

          scaffold/adaptor protein

          Gene Ontology Categories:

          Function(s) Process(es)

          calcium ion transport
          response to cold
          ion transmembrane transport
          response to drug
          response to hydrogen peroxide
          response to pain
          thermoception
          detection of mechanical stimulus involved in sensory perception of pain
          detection of chemical stimulus involved in sensory perception of pain
          calcium ion transmembrane transport
          ion channel activity
          calcium channel activity
          channel activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P472874 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 124576 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 75120 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 415394 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 675512 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 147701 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 952311 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 857375 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 433315 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 97942 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 814331 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 429661 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 58335 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 245755 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 175778 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 621978 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 1953 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 889087 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 550762 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 49905 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 545775 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 644510 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 257778 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 903296 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 674635 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 45410 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 732415 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 187415 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 13464 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 328015 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 765841 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 437604 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 683971 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 99533 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 712523 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 482374 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 91924 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 14868 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 66325 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 47660 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 31446 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 79212 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P472874 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P472874 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P472874 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P472874 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 54402 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 58886 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 49843 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 22802 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10867112 - -
          Affymetrix Rat Gene 1.0 ST Array 10867115 - -
          Affymetrix Rat Gene 1.0 ST Array 10867124 - -
          Affymetrix Rat Gene 1.0 ST Array 10867120 - -
          Affymetrix Rat Gene 1.0 ST Array 10867132 - -
          Affymetrix Rat Gene 1.0 ST Array 10867129 - -
          Affymetrix Rat Gene 1.0 ST Array 10867122 - -
          Affymetrix Rat Gene 1.0 ST Array 10867113 - -
          Affymetrix Rat Gene 1.0 ST Array 10867135 - -
          Affymetrix Rat Gene 1.0 ST Array 10867125 - -
          Affymetrix Rat Gene 1.0 ST Array 10867127 - -
          Affymetrix Rat Gene 1.0 ST Array 10867131 - -
          Affymetrix Rat Gene 1.0 ST Array 10867119 - -
          Affymetrix Rat Gene 1.0 ST Array 10867118 - -
          Affymetrix Rat Gene 1.0 ST Array 10867134 - -
          Affymetrix Rat Gene 1.0 ST Array 10867137 - -
          Affymetrix Rat Gene 1.0 ST Array 10867121 - -
          Affymetrix Rat Gene 1.0 ST Array 10867133 - -
          Affymetrix Rat Gene 1.0 ST Array 10867138 - -
          Affymetrix Rat Gene 1.0 ST Array 10867130 - -
          Affymetrix Rat Gene 1.0 ST Array 10867116 - -
          Affymetrix Rat Gene 1.0 ST Array 10867128 - -
          Affymetrix Rat Gene 1.0 ST Array 10867123 - -
          Affymetrix Rat Gene 1.0 ST Array 10867126 - -
          Affymetrix Rat Gene 1.0 ST Array 10867117 - -
          Affymetrix Rat Gene 1.0 ST Array 10867114 - -
          Affymetrix Rat Gene 1.1 ST Array 10867111 - -
          Affymetrix Rat Clariom S Assay PSR0500000176.rn.1 TC0500000036.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500000177.rn.1 TC0500000036.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500000183.rn.1 TC0500000036.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500000194.rn.1 TC0500000036.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500000195.rn.1 TC0500000036.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500000171.rn.1 TC0500000036.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500000172.rn.1 TC0500000036.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500000173.rn.1 TC0500000036.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500000180.rn.1 TC0500000036.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500000187.rn.1 TC0500000036.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500000188.rn.1 TC0500000036.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500000174.rn.1 TC0500000036.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500000178.rn.1 TC0500000036.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500000181.rn.1 TC0500000036.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500000191.rn.1 TC0500000036.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500000186.rn.1 TC0500000036.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500000189.rn.1 TC0500000036.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500000192.rn.1 TC0500000036.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500000193.rn.1 TC0500000036.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500000179.rn.1 TC0500000036.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500000182.rn.1 TC0500000036.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500000184.rn.1 TC0500000036.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500000185.rn.1 TC0500000036.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500000175.rn.1 TC0500000036.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500000190.rn.1 TC0500000036.rn.1 RTA TC
          Affymetrix Rat Gene 1.0 ST Array 10867111 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21034165 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1353498 - -
          RatRef-12 Expression BeadChip ILMN_51896 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Quick Order
          • eProcurement
          • Supply Center
          • Order Status
          • Chemicals
          • India Mobile App
          • Government eMarketplace
          Support Plus Icon Minus Icon
          • Order Support
          • Training
          • Contact Us
          • Report a Site Issue
          • Instrument Management
          Resources Plus Icon Minus Icon
          • Product Selection Guides
          • Mobile & Desktop Apps
          • Webinars
          • Blog 
          • Social Media
          • New Products
          • Promotions
          • Shared Lists
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          India flag icon
          India

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-jwmpg:80/100.66.76.145:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0