Search
Search

RGD1566265| Gene Symbol: RGD1566265 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001134589.1 | NP_001128061.1 | 2 - 3 | 279 | 103 | |
| GenBank mRNA | FQ229129.1 | - | 2 - 3 | 293 | 103 | |
Set Membership: |
|