Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01474223_g1
          See other DMRTC1C1 GE Assays ›
          Gene Symbol:
          Dmrtc1c1
          Gene Name:
          DMRT-like family C1c1
          Gene Aliases:
          Dmrtc1c, Dmrtc1c2
          Chromosome Location:
          Chr.X: 72684315 - 72691535 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01474223_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Dmrtc1c1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001014222.2 NP_001014244.2 3 - 4 471 81
          GenBank mRNA BC079417.1 - 3 - 4 457 7113562 81
          CV112810.1 - 3 - 4 471 81

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0X00000643.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Dmrtc1c1

          Panther Classification:

          Molecular Function -

          DNA-binding transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of transcription, DNA-templated
          sex differentiation
          biological_process
          core promoter proximal region sequence-specific DNA binding
          molecular_function
          transcription factor activity, sequence-specific DNA binding
          protein homodimerization activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P852728 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 504874 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 839025 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 228956 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 671333 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 730368 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 278616 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 14716 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P852728 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P852728 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P852728 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P852728 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 34546 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10938782 - -
          Affymetrix Rat Gene 1.0 ST Array 10938786 - -
          Affymetrix Rat Gene 1.0 ST Array 10938787 - -
          Affymetrix Rat Gene 1.0 ST Array 10938788 - -
          Affymetrix Rat Gene 1.0 ST Array 10938784 - -
          Affymetrix Rat Gene 1.0 ST Array 10938783 - -
          Affymetrix Rat Gene 1.0 ST Array 10938785 - -
          Affymetrix Rat Gene 1.0 ST Array 10938781 - -
          Affymetrix Rat Gene 1.0 ST Array 10938789 - -
          Affymetrix Rat Gene 1.1 ST Array 10938780 - -
          Rat Genome 230 2.0 Array 1381251_at - -
          Rat Genome 230 2.0 Array 1381251_PM_at - -
          Affymetrix HT Rat Focus Array 1381251_PM_at - -
          Affymetrix HT Rat Focus Array 1381251_at - -
          Affymetrix Rat Clariom S Assay PSR0X00004180.rn.1 TC0X00000643.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0X00004181.rn.1 TC0X00000643.rn.1 RTA TC
          Rat Expression Set 230 1381251_PM_at - -
          Rat Expression Set 230 1381251_at - -
          Affymetrix Rat Gene 1.0 ST Array 10938780 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21241438 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1368104 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-j77tp:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline