Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01474324_m1
          See other APOOL GE Assays ›
          Gene Symbol:
          Apool
          Gene Name:
          apolipoprotein O-like
          Gene Aliases:
          RGD1549725
          Chromosome Location:
          Chr.X: 83678275 - 83750550 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01474324_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Apool
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001014105.1 NP_001014127.1 5 - 6 526 73
          XM_006257191.2 XP_006257253.1 5 - 6 465 73
          GenBank mRNA BC086434.1 - 5 - 6 526 7302620 73
          FQ219134.1 - 5 - 6 416 73
          FQ235331.1 - 5 - 6 412 73

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Apool

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P589570 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 868682 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 109704 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 396177 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 706355 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 574426 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 82558 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 351424 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 250344 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 380483 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 501211 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 755198 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 455523 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 718572 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 34483 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 744912 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 368224 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 98038 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 139184 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 506036 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 340090 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 496425 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 453921 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 676195 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 477454 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 718194 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 879781 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 73947 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 113656 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 617546 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 791238 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 269787 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 632756 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 283560 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 244813 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 879999 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 709811 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 15190 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 124366 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 233286 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 137889 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 916855 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 140071 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 79087 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 120229 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 110799 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 32941 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 152380 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 146197 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P589570 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P589570 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P589570 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P17458 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P589570 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 66490 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 49279 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 1425 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 25871 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 1251 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10934708 - -
          Affymetrix Rat Gene 1.0 ST Array 10934704 - -
          Affymetrix Rat Gene 1.0 ST Array 10934707 - -
          Affymetrix Rat Gene 1.0 ST Array 10934705 - -
          Affymetrix Rat Gene 1.0 ST Array 10934702 - -
          Affymetrix Rat Gene 1.0 ST Array 10934706 - -
          Affymetrix Rat Gene 1.0 ST Array 10934709 - -
          Affymetrix Rat Gene 1.0 ST Array 10934703 - -
          Affymetrix Rat Gene 1.0 ST Array 10934701 - -
          Affymetrix Rat Gene 1.1 ST Array 10934700 - -
          Rat Genome 230 2.0 Array 1389148_at - -
          Rat Genome 230 2.0 Array 1389148_PM_at - -
          Affymetrix HT Rat Focus Array 1389148_at - -
          Affymetrix HT Rat Focus Array 1389148_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0X00004797.rn.1 TC0X00000774.rn.1 RTA TC
          Rat Expression Set 230 1389148_at - -
          Rat Expression Set 230 1389148_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10934700 - -
          Rat Genome U34 Set rc_AI177050_at - -
          Rat Genome U34 Set rc_AI104883_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20836779 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1369989 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-6wvps:80/100.66.75.98:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0