Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01474352_s1
          See other NAP1L2 GE Assays ›
          Gene Symbol:
          Nap1l2
          Gene Name:
          nucleosome assembly protein 1-like 2
          Gene Aliases:
          -
          Chromosome Location:
          Chr.X: 73997274 - 73999677 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01474352_s1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Nap1l2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001024789.1 NP_001019960.1 1 1963 167
          GenBank mRNA BC094525.1 - 1 1963 7318126 167

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Nap1l2

          Panther Classification:

          Molecular Function -

          chromatin/chromatin-binding, or -regulatory protein

          Gene Ontology Categories:

          Function(s) Process(es)

          positive regulation of histone acetylation
          positive regulation of neuron differentiation
          positive regulation of histone H3-K14 acetylation
          regulation of stem cell division
          positive regulation of histone H3-K9 acetylation
          chromatin binding
          histone binding

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P241439 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 506169 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 47370 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 33702 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P241439 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P241439 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P241439 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P241439 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 81848 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10938809 - -
          Affymetrix Rat Gene 1.0 ST Array 10938808 - -
          Affymetrix Rat Gene 1.0 ST Array 10938807 - -
          Affymetrix Rat Gene 1.1 ST Array 10938806 - -
          Rat Genome 230 2.0 Array 1383673_at - -
          Rat Genome 230 2.0 Array 1383673_PM_at - -
          Affymetrix HT Rat Focus Array 1383673_PM_at - -
          Affymetrix HT Rat Focus Array 1383673_at - -
          Affymetrix Rat Clariom S Assay PSR0X00012667.rn.1 TC0X00001999.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0X00012665.rn.1 TC0X00001999.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0X00012666.rn.1 TC0X00001999.rn.1 RTA TC
          Rat Expression Set 230 1383673_at - -
          Rat Expression Set 230 1383673_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10938806 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20794894 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline