Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01474603_m1
          See other NFKBIZ GE Assays ›
          Gene Symbol:
          Nfkbiz
          Gene Name:
          nuclear factor of kappa light polypeptide gene enhancer in B-cells inhibitor, zeta
          Gene Aliases:
          Mail, RGD1310834
          Chromosome Location:
          Chr.11: 47243342 - 47271487 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01474603_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Nfkbiz
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001107095.1 NP_001100565.1 9 - 10 1617 84

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1100000492.rn.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          protein-binding activity modulator

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of transcription from RNA polymerase II promoter
          inflammatory response
          transcription cofactor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P359684 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 725893 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 739554 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 884209 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 397236 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 185354 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 865579 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 805469 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 636196 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 619658 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 49646 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 590718 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 211003 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 606302 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 341085 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 704423 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 800049 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 72293 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 839367 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 964604 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 930588 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 448687 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 822808 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 597044 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 159668 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 39644 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 47871 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 36209 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P359684 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P359684 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P359684 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P19332 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P359684 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 49030 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10750850 - -
          Affymetrix Rat Gene 1.0 ST Array 10750849 - -
          Affymetrix Rat Gene 1.0 ST Array 10750851 - -
          Affymetrix Rat Gene 1.0 ST Array 10750860 - -
          Affymetrix Rat Gene 1.0 ST Array 10750857 - -
          Affymetrix Rat Gene 1.0 ST Array 10750856 - -
          Affymetrix Rat Gene 1.0 ST Array 10750858 - -
          Affymetrix Rat Gene 1.0 ST Array 10750855 - -
          Affymetrix Rat Gene 1.0 ST Array 10750859 - -
          Affymetrix Rat Gene 1.1 ST Array 10750848 - -
          Rat Genome 230 2.0 Array 1378032_at - -
          Rat Genome 230 2.0 Array 1380798_PM_at - -
          Rat Genome 230 2.0 Array 1378032_PM_at - -
          Rat Genome 230 2.0 Array 1380798_at - -
          Affymetrix HT Rat Focus Array 1378032_at - -
          Affymetrix HT Rat Focus Array 1378032_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1100002528.rn.1 TC1100000492.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1100002529.rn.1 TC1100000492.rn.1 RTA TC
          Rat Expression Set 230 1378032_PM_at - -
          Rat Expression Set 230 1378032_at - -
          Rat Expression Set 230 1380798_PM_at - -
          Rat Expression Set 230 1380798_at - -
          Affymetrix Rat Gene 1.0 ST Array 10750848 - -
          Rat Genome U34 Set rc_AI176265_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21314129 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1362295 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-mrft7:80/100.66.76.145:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0