Search
Search

RGD1310935| Gene Symbol: RGD1310935 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008768658.1 | XP_008766880.1 | 6 - 7 | 1584 | 135 | |
| XM_008768659.1 | XP_008766881.1 | 8 - 9 | 1143 | 135 | ||
| XM_008768660.1 | XP_008766882.1 | 7 - 8 | 1500 | 135 | ||
| XM_008768661.1 | XP_008766883.1 | 6 - 7 | 1554 | 135 | ||
| GenBank mRNA | FQ229246.1 | - | 4 - 5 | 615 | 135 | |
Set Membership: |
|