Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01474649_m1
          See other UPK1B GE Assays ›
          Gene Symbol:
          Upk1b
          Gene Name:
          uroplakin 1B
          Gene Aliases:
          -
          Chromosome Location:
          Chr.11: 64522008 - 64553234 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01474649_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Upk1b
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001024253.1 NP_001019424.1 5 - 6 522 68
          XM_006248367.2 XP_006248429.1 5 - 6 729 68
          XM_006248368.2 XP_006248430.1 5 - 6 569 68
          GenBank mRNA BC093613.1 - 5 - 6 522 7309649 68

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1100000629.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Upk1b

          Gene Ontology Categories:

          Function(s)

          epithelial cell differentiation

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P11086 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P130269 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 360881 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 677792 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 855008 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 126904 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 226186 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 318406 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 669430 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 959445 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 691649 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 935227 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 320824 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 783471 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 432075 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 579310 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 555486 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 740236 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 705671 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 128453 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 568586 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 333750 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 447215 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 66441 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 117114 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 11286 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 144232 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P11086 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P130269 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P11086 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P130269 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P130269 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P11086 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P11086 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P384974 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 33610 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 58621 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 65252 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10751248 - -
          Affymetrix Rat Gene 1.0 ST Array 10751245 - -
          Affymetrix Rat Gene 1.0 ST Array 10751247 - -
          Affymetrix Rat Gene 1.0 ST Array 10751250 - -
          Affymetrix Rat Gene 1.0 ST Array 10751252 - -
          Affymetrix Rat Gene 1.0 ST Array 10751249 - -
          Affymetrix Rat Gene 1.0 ST Array 10751253 - -
          Affymetrix Rat Gene 1.0 ST Array 10751251 - -
          Affymetrix Rat Gene 1.0 ST Array 10751254 - -
          Affymetrix Rat Gene 1.1 ST Array 10751239 - -
          Rat Genome 230 2.0 Array 1383875_PM_at - -
          Rat Genome 230 2.0 Array 1383875_at - -
          Affymetrix HT Rat Focus Array 1383875_PM_at - -
          Affymetrix HT Rat Focus Array 1383875_at - -
          Affymetrix Rat Clariom S Assay PSR1100003358.rn.1 TC1100000629.rn.1 RTA TC
          Rat Expression Set 230 1383875_at - -
          Rat Expression Set 230 1383875_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10751239 - -
          Rat Genome U34 Set rc_AI169051_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21269760 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1360729 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-h9fps:80/100.66.77.141:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline