Search
Search
| Gene Symbol: Qtrtd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006248320.2 | XP_006248382.1 | 9 - 10 | 1282 | 91 | |
| XM_008768759.1 | XP_008766981.1 | 9 - 10 | 1321 | 91 | ||
| XM_008768760.1 | XP_008766982.1 | 8 - 9 | 1108 | 91 | ||
| XM_008768761.1 | XP_008766983.1 | 8 - 9 | 1069 | 91 | ||
| XM_008768762.1 | XP_008766984.1 | 7 - 8 | 939 | 91 | ||
Set Membership: |
|