Search
Search
| Gene Symbol: Abi3bp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006248232.2 | XP_006248294.1 | 35 - 36 | 2847 | 82 | |
| XM_006248233.2 | XP_006248295.1 | 28 - 29 | 2420 | 82 | ||
| XM_008768665.1 | XP_008766887.1 | 36 - 37 | 2928 | 82 | ||
| XM_008768666.1 | XP_008766888.1 | 36 - 37 | 2883 | 82 | ||
| XM_008768667.1 | XP_008766889.1 | 32 - 33 | 2654 | 82 | ||
| XM_008768668.1 | XP_008766890.1 | 31 - 32 | 2591 | 82 | ||
| XM_008768669.1 | XP_008766891.1 | 29 - 30 | 2462 | 82 | ||
Set Membership: |
|