Search
Search
| Gene Symbol: Thoc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001191756.1 | NP_001178685.1 | 33 - 34 | 4340 | 69 | |
| XM_006257504.2 | XP_006257566.1 | 33 - 34 | 4432 | 69 | ||
| XM_006257505.2 | XP_006257567.1 | 33 - 34 | 4351 | 69 | ||
Set Membership: |
|