Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01475705_m1
          See other CCDC114 GE Assays ›
          Gene Symbol:
          Ccdc114
          Gene Name:
          coiled-coil domain containing 114
          Gene Aliases:
          RGD1308141
          Chromosome Location:
          Chr.1: 101904047 - 101932999 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01475705_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ccdc114
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001126277.2 NP_001119749.1 11 - 12 1412 131
          XM_008759387.1 XP_008757609.1 12 - 13 3328 131
          XM_008759388.1 XP_008757610.1 11 - 12 1664 131
          XM_008759389.1 XP_008757611.1 8 - 9 986 131
          GenBank mRNA BC160835.1 - 11 - 12 1414 7456475 131

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100001519.rn.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Probe spans exons

          Gene Ontology Categories:

          Function(s)

          outer dynein arm assembly

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P412373 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 208996 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 656718 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 153273 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 311937 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 555036 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 233933 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 340821 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 302132 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 499168 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 589328 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 894289 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 70224 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 443427 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 718124 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 560354 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 544155 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 638031 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 94372 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 533 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 143187 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P412373 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P412373 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P412373 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P412373 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 14111 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10707018 - -
          Affymetrix Rat Gene 1.0 ST Array 10707015 - -
          Affymetrix Rat Gene 1.0 ST Array 10707016 - -
          Affymetrix Rat Gene 1.0 ST Array 10707006 - -
          Affymetrix Rat Gene 1.0 ST Array 10707007 - -
          Affymetrix Rat Gene 1.0 ST Array 10707014 - -
          Affymetrix Rat Gene 1.0 ST Array 10707013 - -
          Affymetrix Rat Gene 1.0 ST Array 10707011 - -
          Affymetrix Rat Gene 1.0 ST Array 10707008 - -
          Affymetrix Rat Gene 1.0 ST Array 10707004 - -
          Affymetrix Rat Gene 1.0 ST Array 10707017 - -
          Affymetrix Rat Gene 1.0 ST Array 10707010 - -
          Affymetrix Rat Gene 1.0 ST Array 10707005 - -
          Affymetrix Rat Gene 1.0 ST Array 10707009 - -
          Affymetrix Rat Gene 1.0 ST Array 10707012 - -
          Affymetrix Rat Gene 1.1 ST Array 10707003 - -
          Affymetrix Rat Clariom S Assay PSR0100012184.rn.1 TC0100001519.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100012182.rn.1 TC0100001519.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100012181.rn.1 TC0100001519.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100012183.rn.1 TC0100001519.rn.1 RTA TC
          Affymetrix Rat Gene 1.0 ST Array 10707003 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21684579 - -
          ABI Rat Genome Survey Microarray 21922009 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1367525 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline