Search
Search
| Gene Symbol: Tex101 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_139037.1 | NP_620606.1 | 1 - 2 | 65 | 83 | |
| XM_006228381.2 | XP_006228443.1 | 2 - 3 | 229 | 83 | ||
| GenBank mRNA | AF347056.1 | - | 1 - 2 | 65 | 83 | |
Set Membership: |
|