Search
Search
| Gene Symbol: Nav2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_138529.1 | NP_612538.1 | 34 - 35 | 6633 | 80 | |
| XM_006229244.2 | XP_006229306.1 | 36 - 37 | 7154 | 80 | ||
| XM_008759312.1 | XP_008757534.1 | 35 - 36 | 7664 | 80 | ||
| XM_008759313.1 | XP_008757535.1 | 34 - 35 | 7655 | 80 | ||
| XM_008759314.1 | XP_008757536.1 | 34 - 35 | 6756 | 80 | ||
| XM_008759315.1 | XP_008757537.1 | 30 - 31 | 6982 | 80 | ||
Set Membership: |
|