Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01475930_m1
          See other FAM98C GE Assays ›
          Gene Symbol:
          Fam98c
          Gene Name:
          family with sequence similarity 98, member C
          Gene Aliases:
          RGD1303117
          Chromosome Location:
          Chr.1: 88095240 - 88098816 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01475930_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Fam98c
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001198584.1 NP_001185513.1 6 - 7 750 94
          NR_037147.1 5 - 6 825 94
          XM_006228717.2 XP_006228779.1 6 - 7 780 94
          GenBank mRNA BC079080.1 - 5 - 6 788 7122702 94
          CO564588.1 - 4 - 5 527 94

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100005118.rn.1

          Set Membership:

          Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          RNA metabolism protein RNA processing factor RNA splicing factor nucleic acid metabolism protein

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P997962 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 759191 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 741761 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 88402 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P997962 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P997962 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P997962 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P997962 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 90378 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10720420 - -
          Affymetrix Rat Gene 1.0 ST Array 10720421 - -
          Affymetrix Rat Gene 1.0 ST Array 10705730 - -
          Affymetrix Rat Gene 1.0 ST Array 10720423 - -
          Affymetrix Rat Gene 1.0 ST Array 10720419 - -
          Affymetrix Rat Gene 1.0 ST Array 10720416 - -
          Affymetrix Rat Gene 1.0 ST Array 10720417 - -
          Affymetrix Rat Gene 1.0 ST Array 10720422 - -
          Affymetrix Rat Gene 1.0 ST Array 10720418 - -
          Affymetrix Rat Gene 1.1 ST Array 10720415 - -
          Rat Genome 230 2.0 Array 1380037_at - -
          Rat Genome 230 2.0 Array 1380037_PM_at - -
          Rat Genome 230 2.0 Array 1377214_a_at - -
          Rat Genome 230 2.0 Array 1377214_PM_a_at - -
          Affymetrix HT Rat Focus Array 1377214_PM_a_at - -
          Affymetrix HT Rat Focus Array 1377214_a_at - -
          Affymetrix Rat Clariom S Assay PSR0100040923.rn.1 TC0100005118.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100040925.rn.1 TC0100005118.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100040928.rn.1 TC0100005118.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100040924.rn.1 TC0100005118.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100040926.rn.1 TC0100005118.rn.1 RTA TC
          Rat Expression Set 230 1377214_PM_a_at - -
          Rat Expression Set 230 1377214_a_at - -
          Affymetrix Rat Gene 1.0 ST Array 10720415 - -
          Rat Genome U34 Set rc_AI070824_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21336058 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1373590 - -
          RatRef-12 Expression BeadChip ILMN_49971 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline