Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01476079_g1
          See other GNG8 GE Assays ›
          Gene Symbol:
          Gng8
          Gene Name:
          guanine nucleotide binding protein (G protein), gamma 8
          Gene Aliases:
          -
          Chromosome Location:
          Chr.1: 78818377 - 78822224 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01476079_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gng8
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_139185.1 NP_631924.1 3 - 4 302 86
          XM_006228384.2 XP_006228446.1 2 - 3 656 86
          XM_006228385.2 XP_006228447.1 2 - 3 100 86
          GenBank mRNA L35921.1 - 3 - 4 302 86

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100000942.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Gng8

          Panther Classification:

          Molecular Function -

          heterotrimeric G-protein

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          nervous system development
          social behavior
          nose development
          cellular response to pheromone
          signal transducer activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P529550 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 70273 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 812408 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P529550 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P529550 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P529550 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P529550 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10704564 - -
          Affymetrix Rat Gene 1.0 ST Array 10704563 - -
          Affymetrix Rat Gene 1.0 ST Array 10704562 - -
          Affymetrix Rat Gene 1.0 ST Array 10704561 - -
          Affymetrix Rat Gene 1.1 ST Array 10704560 - -
          Rat Genome 230 2.0 Array 1370843_PM_at - -
          Rat Genome 230 2.0 Array 1370843_at - -
          Affymetrix HT Rat Focus Array 1370843_at - -
          Affymetrix HT Rat Focus Array 1370843_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0100006558.rn.1 TC0100000942.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100006559.rn.1 TC0100000942.rn.1 RTA TC
          Rat Expression Set 230 1370843_PM_at - -
          Rat Expression Set 230 1370843_at - -
          Affymetrix Rat Gene 1.0 ST Array 10704560 - -
          Rat Genome U34 Set L35921_g_at - -
          Rat Genome U34 Set L35921_at - -
          Rat Genome U34 Set rc_AI009235_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21307077 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1650457 - -
          RatRef-12 Expression BeadChip ILMN_55153 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-green-7b6576f7bc-knt4q:80/100.66.75.14:80.
          git-commit: 6d309ebe7a31b2ceadeb87f43b473eebab110cf3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-2026.03.11-1.0