Search
Search

Exoc3l2| Gene Symbol: Exoc3l2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006228497.2 | XP_006228559.1 | 10 - 11 | 2048 | 59 | |
| XM_006228498.2 | XP_006228560.1 | 10 - 11 | 2184 | 59 | ||
Set Membership: |
|