Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01476589_m1
          See other PEPD GE Assays ›
          Gene Symbol:
          Pepd
          Gene Name:
          peptidase D
          Gene Aliases:
          -
          Chromosome Location:
          Chr.1: 90820670 - 91285128 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01476589_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Pepd
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001009641.1 NP_001009641.1 13 - 14 1167 96
          GenBank mRNA BC088440.1 - 13 - 14 1167 7126171 96
          FQ235015.1 - 2 - 3 229 96

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100001294.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Protease Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Pepd

          Panther Classification:

          Molecular Function -

          metalloprotease

          Gene Ontology Categories:

          Function(s) Process(es)

          proteolysis
          collagen catabolic process
          aminopeptidase activity
          peptidase activity
          metallopeptidase activity
          dipeptidase activity
          manganese ion binding
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P135604 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 306331 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 129182 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 261453 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 956383 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 480608 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 131440 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 947669 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 751905 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 451409 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 629846 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 671064 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 119609 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 851603 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 485244 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 807621 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 515255 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 517310 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 265085 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 577111 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 914803 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 916677 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 132587 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 30762 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 170242 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 642611 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 905610 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 626913 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 24995 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 132356 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 578509 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 539541 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 663135 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 335924 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 119146 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 214918 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 745912 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 852733 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 573147 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 896562 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 909875 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 614702 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 261315 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 531576 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 463314 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 775202 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 298031 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 717677 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 63273 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 456256 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 466421 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 656315 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 446602 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 337783 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 534260 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 705574 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 911947 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 794351 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 754101 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 98980 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 242758 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 335011 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 26573 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 906609 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 930882 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 764533 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 897034 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 308189 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 233437 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 207557 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 900117 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 775055 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 361957 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 17492 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 75685 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 835805 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 965049 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 40996 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 63017 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 260600 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 583932 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 111899 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 949018 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 43913 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 87661 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 80143 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 50360 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 34529 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 138454 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 25615 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 154511 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 111250 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 154218 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 92256 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 155310 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 56531 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 4149 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 105551 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 132831 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P135604 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P135604 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P135604 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P20408 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 42469 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 81906 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 8367 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 26745 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 44417 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 72194 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 102554 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 27207 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10706129 - -
          Affymetrix Rat Gene 1.0 ST Array 10706122 - -
          Affymetrix Rat Gene 1.0 ST Array 10706121 - -
          Affymetrix Rat Gene 1.0 ST Array 10706128 - -
          Affymetrix Rat Gene 1.0 ST Array 10706118 - -
          Affymetrix Rat Gene 1.0 ST Array 10706123 - -
          Affymetrix Rat Gene 1.0 ST Array 10706130 - -
          Affymetrix Rat Gene 1.0 ST Array 10706120 - -
          Affymetrix Rat Gene 1.0 ST Array 10706119 - -
          Affymetrix Rat Gene 1.0 ST Array 10706126 - -
          Affymetrix Rat Gene 1.0 ST Array 10706124 - -
          Affymetrix Rat Gene 1.0 ST Array 10706127 - -
          Affymetrix Rat Gene 1.0 ST Array 10706125 - -
          Affymetrix Rat Gene 1.1 ST Array 10706117 - -
          Rat Genome 230 2.0 Array 1370699_a_at - -
          Rat Genome 230 2.0 Array 1370699_PM_a_at - -
          Affymetrix Rat Clariom S Assay PSR0100010033.rn.1 TC0100001294.rn.1 RTA TC
          Rat Expression Set 230 1370699_PM_a_at - -
          Rat Expression Set 230 1370699_a_at - -
          Affymetrix Rat Gene 1.0 ST Array 10706117 - -
          Rat Genome U34 Set rc_AA899762_g_at - -
          Rat Genome U34 Set rc_AA899762_at - -
          Rat Genome U34 Set rc_AI233224_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22059436 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1352307 - -
          RatRef-12 Expression BeadChip ILMN_62988 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-hznln:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline