Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01476722_m1
          See other VSTM2B GE Assays ›
          Gene Symbol:
          Vstm2b
          Gene Name:
          V-set and transmembrane domain containing 2B
          Gene Aliases:
          RGD1306235
          Chromosome Location:
          Chr.1: 97632473 - 97661541 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01476722_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Vstm2b
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001108479.1 NP_001101949.1 5 - 6 1372 110
          XM_008759395.1 XP_008757617.1 3 - 4 767 110

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100001385.rn.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Recommended Assay Rattus norvegicus 1700 Probe spans exons

          Panther Classification:

          Molecular Function -

          immunoglobulin receptor superfamily

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P488000 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 823895 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 426543 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 29167 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 620916 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 185872 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 323748 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 632385 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 584049 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 361897 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 805245 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 673448 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 32268 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 366288 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 457325 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 963005 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 470754 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 950011 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 390849 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 65970 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 132688 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 32856 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P488000 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P488000 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P488000 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P488000 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 78638 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10706255 - -
          Affymetrix Rat Gene 1.0 ST Array 10706254 - -
          Affymetrix Rat Gene 1.0 ST Array 10706253 - -
          Affymetrix Rat Gene 1.0 ST Array 10706256 - -
          Affymetrix Rat Gene 1.0 ST Array 10706257 - -
          Affymetrix Rat Gene 1.1 ST Array 10706249 - -
          Rat Genome 230 2.0 Array 1397861_at - -
          Rat Genome 230 2.0 Array 1397861_PM_at - -
          Affymetrix HT Rat Focus Array 1397861_PM_at - -
          Affymetrix HT Rat Focus Array 1397861_at - -
          Affymetrix Rat Clariom S Assay PSR0100010610.rn.1 TC0100001385.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100010611.rn.1 TC0100001385.rn.1 RTA TC
          Rat Expression Set 230 1397861_at - -
          Rat Expression Set 230 1397861_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10706249 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21821439 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1366621 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-kj7pn:80/100.66.78.100:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0